Search
Search

| Gene Symbol: Olr1525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000529.1 | NP_001000529.1 | 919 | |||
| XM_008767530.1 | XP_008765752.1 | 1 | 919 | |||
| Gene Symbol: Olr1355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214823.1 | NP_999988.1 | 1 | 919 | ||