Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LDB2 SIRNA Assays ›
Gene Symbol:
LDB2
Gene Name:
LIM domain binding 2
Gene Aliases:
CLIM1, LDB-2, LDB1
Chromosome Location:
Chr.4: 16501534 - 16898809 on Build GRCh38
UniGene ID:
Hs.714330
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17322
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LDB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130834.2
NP_001124306.1
4
806
NM_001290.4
NP_001281.1
4
806
NM_001304434.1
NP_001291363.1
4
806
NM_001304435.1
NP_001291364.1
4
806
XM_005248197.3
XP_005248254.1
4
776
XM_005248201.2
XP_005248258.1
4
776
XM_005248202.2
XP_005248259.1
4
776
XM_006713975.3
XP_006714038.1
6
835
XM_006713976.3
XP_006714039.1
6
836
XM_006713977.3
XP_006714040.1
6
835
XM_006713978.3
XP_006714041.1
6
836
XM_006713979.3
XP_006714042.1
6
836
XM_006713980.3
XP_006714043.1
6
837
XM_006713981.3
XP_006714044.1
6
836
XM_006713982.3
XP_006714045.1
6
836
XM_006713983.3
XP_006714046.1
6
836
XM_011513905.2
XP_011512207.1
5
727
XM_017008812.1
XP_016864301.1
6
836
XM_017008813.1
XP_016864302.1
4
776
XM_017008814.1
XP_016864303.1
6
837
XM_017008815.1
XP_016864304.1
6
837
XM_017008816.1
XP_016864305.1
6
837
XM_017008817.1
XP_016864306.1
4
776
XM_017008818.1
XP_016864307.1
4
373
XM_017008819.1
XP_016864308.1
4
776
XM_017008820.1
XP_016864309.1
4
776
XM_017008821.1
XP_016864310.1
4
776
XM_017008822.1
XP_016864311.1
5
716
GenBank mRNA
AF047337.1
-
4
591
AF052389.1
-
5
664
AF064492.1
-
4
591
AF064493.1
-
4
591
AF068651.1
-
4
589
AK226029.1
-
4
631
AK294588.1
-
3
361
AK300119.1
-
4
403
AK312881.1
-
4
501
BC034019.1
-
4
599
4693654
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
hair follicle development
epithelial structure maintenance
regulation of cell migration
somatic stem cell population maintenance
regulation of kinase activity
positive regulation of cellular component biogenesis
positive regulation of transcription from RNA polymerase II promoter
transcription cofactor activity
enzyme binding
LIM domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-s9xdw:80/100.66.76.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0