Search
Search

SLC16A3| Gene Symbol: SLC16A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042422.2 | NP_001035887.1 | 2 | 295 | ||
| NM_001042423.2 | NP_001035888.1 | 2 | 299 | |||
| NM_001206950.1 | NP_001193879.1 | 2 | 311 | |||
| NM_001206951.1 | NP_001193880.1 | 2 | 273 | |||
| NM_001206952.1 | NP_001193881.1 | 2 | 240 | |||
| NM_004207.3 | NP_004198.1 | 2 | 267 | |||
| XM_011523607.1 | XP_011521909.1 | 2 | 2188 | |||
| GenBank mRNA | AK127319.1 | - | 1 | 2455 | ||
| AK223040.1 | - | 2 | 225 | |||
| BC112267.1 | - | 1 | 186 | 8327735 | ||
| BC112269.1 | - | 1 | 186 | 8327737 | ||
| BM926587.1 | - | 2 | 308 | |||
| CX872345.1 | - | 2 | 299 | |||
| DA373476.1 | - | 2 | 274 | |||
| DA419716.1 | - | 2 | 272 | |||
| DA583988.1 | - | 2 | 296 | |||
| DA895850.1 | - | 2 | 240 | |||
| U81800.1 | - | 2 | 207 | |||