Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP5A1 SIRNA Assays ›
          Gene Symbol:
          ATP5A1
          Gene Name:
          ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle
          Gene Aliases:
          ATP5A, ATP5AL2, ATPM, COXPD22, HEL-S-123m, MC5DN4, MOM2, OMR, ORM, hATP1
          Chromosome Location:
          Chr.18: 46084144 - 46104233 on Build GRCh38
          UniGene ID:
          Hs.298280
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1767
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATP5A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001935.2 NP_001001935.1 5, 6 853
          NM_001001937.1 NP_001001937.1 6, 7 803
          NM_001257334.1 NP_001244263.1 5, 6 714
          NM_001257335.1 NP_001244264.1 5, 6 1116
          NM_004046.5 NP_004037.1 5, 6 780
          XM_017025789.1 XP_016881278.1 5, 6 722
          GenBank mRNA AK092735.1 - 5, 6 795
          AK098765.1 - 5, 6 712
          AK129739.1 - 5, 6 723
          AK129900.1 - 5, 6 723
          AK289457.1 - 5, 6 1049
          AK302272.1 - 5, 6 657
          AU120644.1 - 5, 6 748
          BC003119.1 - 1, 2 103
          BC007299.1 - 2, 3 223
          BC008028.2 - 2, 3 350
          BC011384.1 - 5, 6 711 3867375
          BC016046.1 - 5, 6 698 4646761
          BC019310.1 - 5, 6 711 2823663
          BC028335.1 - 5, 6 700
          BC039135.1 - 5, 6 693 3873390
          BC042694.1 - 5, 6 693
          BC064562.1 - 5, 6 711 5582043
          BC067385.1 - 5, 6 723 5491013
          BT007209.1 - 5, 6 658
          CD244623.1 - 5, 6 739
          D14710.1 - 5, 6 721
          FJ224309.1 - 5, 6 716
          X59066.1 - 5, 6 716
          X65460.1 - 5, 6 672

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00818620_g1

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endothelial cell proliferation
          lipid metabolic process
          ATP biosynthetic process
          embryo development
          ATP hydrolysis coupled proton transport
          mitochondrial ATP synthesis coupled proton transport
          protein binding
          ATP binding
          ATPase activity
          transmembrane transporter activity
          MHC class I protein binding
          poly(A) RNA binding
          proton-transporting ATP synthase activity, rotational mechanism
          proton-transporting ATPase activity, rotational mechanism

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0