Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LILRA3 SIRNA Assays ›
          Gene Symbol:
          Lilra3 LOC100912456 LOC690948
          Gene Name:
          leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 3
          leukocyte immunoglobulin-like receptor subfamily B member 3-like
          similar to paired-Ig-like receptor A11
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.1: 63920125 - 63929449 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 63883682 - 63891050 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 63963853 - 63971222 on Build Rnor_6.0
          UniGene ID:
          -
          -
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s186206

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lilra3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003748712.3 XP_003748760.1 613
          XM_006228047.2 XP_006228109.1 615
          XM_006228049.2 XP_006228111.1 614
          XM_006228050.2 XP_006228112.1 616
          XM_006228052.2 XP_006228114.1 426
          XM_006228053.2 XP_006228115.1 540
          Gene Symbol: LOC100912456
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003748711.3 XP_003748759.1 4 615
          XM_006228039.2 XP_006228101.1 4 616
          XM_006228040.2 XP_006228102.1 4 614
          XM_006228042.2 XP_006228104.1 4 626
          XM_006228043.2 XP_006228105.1 3 541
          Gene Symbol: LOC690948
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_001076276.4 XP_001076276.1 615
          XM_006228055.2 XP_006228117.1 616
          XM_006228056.2 XP_006228118.1 614
          XM_006228058.2 XP_006228120.1 626
          XM_006228059.2 XP_006228121.1 541

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of T cell mediated cytotoxicity
          positive regulation of defense response to virus by host
          T cell proliferation involved in immune response
          negative regulation of cytokine secretion involved in immune response
          Fc receptor mediated inhibitory signaling pathway
          signal transduction
          response to virus
          positive regulation of gene expression
          negative regulation of serotonin secretion
          interferon-gamma production
          negative regulation of interferon-gamma production
          negative regulation of mononuclear cell proliferation
          negative regulation of interferon-beta secretion
          negative regulation of T cell proliferation
          negative regulation of tumor necrosis factor biosynthetic process
          positive regulation of apoptotic process
          negative regulation of interferon-gamma biosynthetic process
          negative regulation of cell cycle
          negative regulation of endocytosis
          positive regulation of cytolysis
          negative regulation of natural killer cell mediated cytotoxicity
          negative regulation of alpha-beta T cell activation
          defense response to virus
          negative regulation of calcium ion transport
          cellular response to lipopolysaccharide
          interferon-gamma secretion
          dendritic cell differentiation
          negative regulation of dendritic cell apoptotic process
          negative regulation of interleukin-10 secretion
          negative regulation of interleukin-12 secretion
          negative regulation of CD8-positive, alpha-beta T cell activation
          negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          positive regulation of gamma-delta T cell activation involved in immune response
          negative regulation of dendritic cell differentiation
          negative regulation of transforming growth factor-beta secretion
          negative regulation of osteoclast development
          protein phosphatase 1 binding
          HLA-A specific inhibitory MHC class I receptor activity
          HLA-B specific inhibitory MHC class I receptor activity
          MHC class I receptor activity
          SH2 domain binding
          MHC class I protein binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0