Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNAP91 SIRNA Assays ›
          Gene Symbol:
          SNAP91
          Gene Name:
          synaptosome associated protein 91
          Gene Aliases:
          AP180, CALM
          Chromosome Location:
          Chr.6: 83552885 - 83709408 on Build GRCh38
          UniGene ID:
          Hs.368046
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s19163

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNAP91
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242792.1 NP_001229721.1 7, 8 965
          NM_001242793.1 NP_001229722.1 7, 8 764
          NM_001242794.1 NP_001229723.1 7, 8 764
          NM_001256717.1 NP_001243646.1 10, 11 1200
          NM_001256718.1 NP_001243647.1 6, 7 675
          NM_014841.2 NP_055656.1 7, 8 968
          XM_005248770.4 XP_005248827.1 7, 8 807
          XM_006715615.1 XP_006715678.1 7, 8 759
          XM_011536265.1 XP_011534567.1 7, 8 766
          XM_011536266.1 XP_011534568.1 7, 8 878
          XM_011536269.1 XP_011534571.1 7, 8 759
          XM_011536271.1 XP_011534573.1 7, 8 759
          XM_011536273.2 XP_011534575.1 7, 8 759
          XM_011536275.2 XP_011534577.1 7, 8 759
          XM_011536276.2 XP_011534578.1 7, 8 759
          XM_011536277.1 XP_011534579.1 9, 10 1047
          XM_017011554.1 XP_016867043.1 7, 8 923
          XM_017011555.1 XP_016867044.1 7, 8 807
          XM_017011556.1 XP_016867045.1 7, 8 739
          XM_017011557.1 XP_016867046.1 7, 8 936
          XM_017011558.1 XP_016867047.1 7, 8 766
          XM_017011559.1 XP_016867048.1 7, 8 865
          XM_017011560.1 XP_016867049.1 7, 8 807
          XM_017011561.1 XP_016867050.1 7, 8 878
          XM_017011562.1 XP_016867051.1 7, 8 936
          XM_017011563.1 XP_016867052.1 7, 8 936
          XM_017011564.1 XP_016867053.1 7, 8 807
          XM_017011565.1 XP_016867054.1 7, 8 865
          XM_017011566.1 XP_016867055.1 7, 8 936
          XM_017011567.1 XP_016867056.1 7, 8 739
          XM_017011568.1 XP_016867057.1 7, 8 936
          XM_017011569.1 XP_016867058.1 7, 8 936
          XM_017011570.1 XP_016867059.1 7, 8 936
          XM_017011571.1 XP_016867060.1 7, 8 807
          XM_017011572.1 XP_016867061.1 7, 8 766
          XM_017011573.1 XP_016867062.1 7, 8 865
          XM_017011574.1 XP_016867063.1 7, 8 936
          XM_017011575.1 XP_016867064.1 7, 8 936
          XM_017011576.1 XP_016867065.1 7, 8 739
          XM_017011577.1 XP_016867066.1 7, 8 865
          XM_017011578.1 XP_016867067.1 7, 8 878
          XM_017011579.1 XP_016867068.1 7, 8 936
          XM_017011580.1 XP_016867069.1 7, 8 936
          XM_017011581.1 XP_016867070.1 7, 8 766
          XM_017011582.1 XP_016867071.1 7, 8 795
          XM_017011583.1 XP_016867072.1 7, 8 923
          XM_017011584.1 XP_016867073.1 7, 8 766
          XM_017011585.1 XP_016867074.1 7, 8 807
          XM_017011586.1 XP_016867075.1 7, 8 759
          XM_017011587.1 XP_016867076.1 7, 8 865
          XM_017011588.1 XP_016867077.1 7, 8 878
          XM_017011589.1 XP_016867078.1 7, 8 936
          XM_017011590.1 XP_016867079.1 7, 8 807
          GenBank mRNA AB014556.1 - 7, 8 894
          AK289582.1 - 7, 8 936
          AK294898.1 - 6, 7 659
          BC060818.1 - 7, 8 748 4827211
          CR749348.1 - 10, 11 1167

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          protein transport
          clathrin coat assembly
          regulation of clathrin-mediated endocytosis
          protein binding
          1-phosphatidylinositol binding
          protein kinase binding
          clathrin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0