Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCL4 SIRNA Assays ›
          Gene Symbol:
          CCL4 CCL4L1 CCL4L2
          Gene Name:
          C-C motif chemokine ligand 4
          C-C motif chemokine ligand 4 like 1
          C-C motif chemokine ligand 4 like 2
          Gene Aliases:
          ACT2, AT744.1, G-26, HC21, LAG-1, LAG1, MIP-1-beta, MIP1B, MIP1B1, SCYA2, SCYA4
          AT744.2, CCL4L, LAG-1, LAG1, MIP-1-beta, SCYA4L, SCYA4L1, SCYA4L2
          AT744.2, CCL4L, SCYA4L, SCYQ4L2
          Chromosome Location:
          Chr.17: 36103827 - 36105621 on Build GRCh38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.17: 36211063 - 36212878 on Build GRCh38
          UniGene ID:
          Hs.75703
          Hs.449862
          Hs.515183
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s194791

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCL4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002984.3 NP_002975.1 3 343
          GenBank mRNA AY312578.1 - 2 160
          AY766446.1 - 3 343
          AY766448.1 - 2 228
          BC104226.1 - 3 334 40032168
          BC104227.1 - 3 334 40032172
          BC107433.1 - 3 344 6727378
          CR542119.1 - 3 264
          J04130.1 - 3 372
          M23502.1 - 3 338
          M25316.1 - 3 337
          M57503.1 - 3 251
          X16166.1 - 3 316
          X53683.1 - 3 330
          Gene Symbol: CCL4L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_207007.3 NP_996890.1 3 498
          NR_111969.1 2 383
          GenBank mRNA AY766447.1 - 3 343
          AY766449.1 - 2 228
          BC070310.1 - 3 340 6708338
          BC130456.1 - 3 305 40146487
          BC130458.1 - 3 305 40146489
          BC144393.1 - 3 305 9052914
          BC144394.1 - 3 305 9052915
          BC146944.1 - 3 498 9056872
          BC146958.1 - 3 498 9056886
          BC171864.1 - 3 498 9054518
          Gene Symbol: CCL4L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291468.1 NP_001278397.1 3 328
          NM_001291469.1 NP_001278398.1 3 532
          NM_001291470.1 NP_001278399.1 2 934
          NM_001291471.1 NP_001278400.1 3 512
          NM_001291472.1 NP_001278401.1 3 491
          NM_001291473.1 NP_001278402.1 3 461
          NM_001291474.1 NP_001278403.1 2 417
          NM_001291475.1 NP_001278404.1 3 522
          NR_111970.1 2 213
          GenBank mRNA AY766450.1 - 3 328
          AY766451.1 - 3 532
          AY766452.1 - 2 934
          AY766453.1 - 3 512
          AY766454.1 - 3 491
          AY766455.1 - 3 461
          AY766456.1 - 2 213
          AY766457.1 - 2 417
          AY766458.1 - 3 522
          BC092445.1 - 3 529 30416435
          CD521054.1 - 1 261

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          monocyte chemotaxis
          movement of cell or subcellular component
          inflammatory response
          immune response
          cell adhesion
          establishment or maintenance of cell polarity
          signal transduction
          G-protein coupled receptor signaling pathway
          cell-cell signaling
          response to virus
          response to toxic substance
          neutrophil chemotaxis
          positive regulation of GTPase activity
          negative regulation by host of viral transcription
          positive regulation of inflammatory response
          positive regulation of calcium-mediated signaling
          positive regulation of calcium ion transport
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          positive regulation of natural killer cell chemotaxis
          lymphocyte chemotaxis
          cytokine activity
          protein binding
          chemokine activity
          CCR1 chemokine receptor binding
          CCR5 chemokine receptor binding
          identical protein binding
          CCR chemokine receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0