Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBMY1E SIRNA Assays ›
          Gene Symbol:
          RBMY1E RBMY1A1 RBMY1A3P RBMY1B RBMY1D RBMY1F RBMY1J RBMY2FP
          Gene Name:
          RNA binding motif protein, Y-linked, family 1, member E
          RNA binding motif protein, Y-linked, family 1, member A1
          RNA binding motif protein, Y-linked, family 1, member A3 pseudogene
          RNA binding motif protein, Y-linked, family 1, member B
          RNA binding motif protein, Y-linked, family 1, member D
          RNA binding motif protein, Y-linked, family 1, member F
          RNA binding motif protein, Y-linked, family 1, member J
          RNA binding motif protein, Y-linked, family 2, member F pseudogene
          Gene Aliases:
          -
          RBM, RBM1, RBM2, RBMY, RBMY1C, YRRM1, YRRM2
          -
          -
          -
          YRRM2
          -
          -
          Chromosome Location:
          Chr.Y: 21902174 - 21918044 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21534879 - 21559683 on Build GRCh38
          Chromosome Location:
          Chr.Y: 9317061 - 9322874 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21511338 - 21527212 on Build GRCh38
          Chromosome Location:
          Chr.Y: 21869719 - 21900746 on Build GRCh38
          Chromosome Location:
          Chr.Y: 22167099 - 22182942 on Build GRCh38
          Chromosome Location:
          Chr.Y: 22403410 - 22419317 on Build GRCh38
          Chromosome Location:
          Chr.Y: 22308859 - 22316205 on Build GRCh38
          UniGene ID:
          Hs.725744
          Hs.380450
          Hs.567833
          Hs.380450
          Hs.380450
          Hs.567746
          Hs.380450
          Hs.711527
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s196168

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBMY1E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006118.2 NP_001006118.2 4 349
          XM_011531493.1 XP_011529795.1 4 349
          XM_011531494.2 XP_011529796.1 4 349
          XM_011531495.1 XP_011529797.1 4 349
          XM_017030058.1 XP_016885547.1 3 165
          Gene Symbol: RBMY1A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320944.1 NP_001307873.1 4 389
          NM_005058.3 NP_005049.1 4 389
          XM_011531437.2 XP_011529739.1 4 328
          XM_011531438.1 XP_011529740.1 4 328
          XM_017030060.1 XP_016885549.1 3 165
          GenBank mRNA BC070298.1 - 4 354 6615329
          BC146853.1 - 4 302 9056781
          BU568309.1 - 4 357
          X76059.1 - 4 354
          Gene Symbol: RBMY1A3P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_001547.1 3 242
          Gene Symbol: RBMY1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006121.3 NP_001006121.1 4 389
          NM_001320948.1 NP_001307877.1 4 389
          XM_011531433.1 XP_011529735.1 4 349
          XM_017030056.1 XP_016885545.1 4 349
          XM_017030057.1 XP_016885546.1 3 165
          GenBank mRNA BC136862.1 - 4 302 9020853
          BC140758.1 - 4 302 9021626
          BC144459.1 - 4 302 9052985
          Gene Symbol: RBMY1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006120.3 NP_001006120.2 4 389
          NM_001320949.1 NP_001307878.1 4 389
          NM_001320950.1 NP_001307879.1 5 440
          XM_011531486.2 XP_011529788.1 4 262
          XM_011531487.2 XP_011529789.1 4 349
          XM_011531488.2 XP_011529790.1 4 349
          XM_011531489.1 XP_011529791.1 4 349
          GenBank mRNA BC144422.1 - 4 302 9052943
          BC144423.1 - 5 353
          Gene Symbol: RBMY1F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303410.1 NP_001290339.1 4 349
          NM_152585.2 NP_689798.1 4 349
          XM_017030033.1 XP_016885522.1 3 165
          GenBank mRNA BC030018.2 - 4 349 5269012
          X76060.1 - 4 331
          Gene Symbol: RBMY1J
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006117.3 NP_001006117.2 4 414
          XM_011531500.1 XP_011529802.1 4 285
          XM_011531502.1 XP_011529804.1 4 285
          XM_017030059.1 XP_016885548.1 3 165
          GenBank mRNA AK313597.1 - 4 418
          BC144577.1 - 4 302 9053106
          BC152392.1 - 3 259 8860211
          Gene Symbol: RBMY2FP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002193.2 4 353
          GenBank mRNA BC026077.2 - 4 355

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          spermatogenesis
          RNA splicing
          male gonad development
          nucleotide binding
          RNA binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0