Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TUBA1B SIRNA Assays ›
          Gene Symbol:
          Tuba1b
          Gene Name:
          tubulin, alpha 1B
          Gene Aliases:
          Tuba2
          Chromosome Location:
          Chr.15: 98931427 - 98934560 on Build GRCm38
          UniGene ID:
          Mm.392113
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s202331

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tuba1b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011654.2 NP_035784.1 4 1408
          GenBank mRNA AK075955.1 - 4 1455
          AK137885.1 - 4 1452
          AK150128.1 - 4 1455
          AK150179.1 - 4 1455
          AK150968.1 - 4 1454
          AK151199.1 - 4 1452
          AK151809.1 - 4 1452
          AK152298.1 - 4 1452
          AK153064.1 - 4 1452
          AK153254.1 - 4 1452
          AK158958.1 - 4 1584
          AK164428.1 - 4 1455
          AK168770.1 - 4 1452
          AK169075.1 - 4 1452
          AK169092.1 - 4 1455
          AK169130.1 - 4 1455
          AK169665.1 - 4 1455
          AK191172.1 - 1 262
          AK192129.1 - 1 241
          AK192775.1 - 1 259
          AK193004.1 - 1 220
          AK194180.1 - 1 220
          AK194222.1 - 1 214
          AK196308.1 - 1 229
          AK197610.1 - 1 262
          AK198269.1 - 1 262
          AK199828.1 - 1 209
          AK200084.1 - 1 262
          AK203041.1 - 1 185
          AK207058.1 - 1 219
          AK211470.1 - 1 230
          BC002219.1 - 4 1442 3490568
          BC004790.1 - 2 1043
          BC008117.1 - 4 1435 3498312
          BC020390.1 - 1295
          BC063777.1 - 4 1405 4458530
          BC083120.1 - 4 1433 6311394
          BC098321.1 - 4 1435 5065638
          BC108337.1 - 4 1434 4167097
          BC108394.1 - 4 1408 6505244
          M13446.1 - 4 1432
          M28727.1 - 2 1056

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          protein polymerization
          cellular response to interleukin-4
          nucleotide binding
          double-stranded RNA binding
          GTPase activity
          structural constituent of cytoskeleton
          protein binding
          GTP binding
          ubiquitin protein ligase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-tc65k:80/100.66.76.115:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0