Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NCOR1 SIRNA Assays ›
          Gene Symbol:
          NCOR1
          Gene Name:
          nuclear receptor corepressor 1
          Gene Aliases:
          N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
          Chromosome Location:
          Chr.17: 16030094 - 16216746 on Build GRCh38
          UniGene ID:
          Hs.462323
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s203
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NCOR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190438.1 NP_001177367.1 17 2355
          NM_001190440.1 NP_001177369.1 19 2682
          NM_006311.3 NP_006302.2 20 2833
          XM_005256866.4 XP_005256923.1 21 2894
          XM_005256868.4 XP_005256925.1 21 2891
          XM_005256871.4 XP_005256928.1 20 2867
          XM_005256872.4 XP_005256929.1 20 2864
          XM_005256873.4 XP_005256930.1 21 2846
          XM_005256874.4 XP_005256931.1 21 2894
          XM_005256875.3 XP_005256932.1 21 2894
          XM_006721601.3 XP_006721664.1 22 3026
          XM_006721602.3 XP_006721665.1 22 3021
          XM_006721603.3 XP_006721666.1 21 2894
          XM_006721604.3 XP_006721667.1 20 2864
          XM_011524083.2 XP_011522385.1 21 3805
          XM_011524084.2 XP_011522386.1 21 2894
          XM_011524085.2 XP_011522387.1 21 2894
          XM_011524086.2 XP_011522388.1 20 2864
          XM_017025396.1 XP_016880885.1 22 3771
          XM_017025397.1 XP_016880886.1 22 3536
          XM_017025398.1 XP_016880887.1 21 2978
          XM_017025399.1 XP_016880888.1 21 2997
          XM_017025400.1 XP_016880889.1 22 3768
          XM_017025401.1 XP_016880890.1 22 3533
          XM_017025402.1 XP_016880891.1 21 2817
          XM_017025403.1 XP_016880892.1 21 2994
          XM_017025404.1 XP_016880893.1 21 3485
          XM_017025405.1 XP_016880894.1 20 3380
          XM_017025406.1 XP_016880895.1 20 3778
          XM_017025407.1 XP_016880896.1 20 2714
          XM_017025408.1 XP_016880897.1 20 2786
          XM_017025409.1 XP_016880898.1 21 2991
          XM_017025410.1 XP_016880899.1 21 3487
          XM_017025411.1 XP_016880900.1 20 3230
          XM_017025412.1 XP_016880901.1 20 3007
          XM_017025413.1 XP_016880902.1 20 2790
          XM_017025414.1 XP_016880903.1 19 2697
          XM_017025415.1 XP_016880904.1 21 2894
          XM_017025416.1 XP_016880905.1 21 3502
          XM_017025417.1 XP_016880906.1 21 2846
          XM_017025418.1 XP_016880907.1 21 2894
          XM_017025419.1 XP_016880908.1 21 2894
          XM_017025420.1 XP_016880909.1 21 2846
          GenBank mRNA AB028970.2 - 19 2656
          AF044209.1 - 20 2804
          AF087856.1 - 17 2369
          AF303586.1 - 17 2369
          BC167431.1 - 19 2657 8860107

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00196920_m1

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          skeletal muscle tissue development
          lactation
          circadian rhythm
          positive regulation of cell proliferation
          covalent chromatin modification
          cerebellum development
          cellular lipid metabolic process
          negative regulation of JNK cascade
          spindle assembly
          regulation of glycolytic process by negative regulation of transcription from RNA polymerase II promoter
          regulation of lipid transport by negative regulation of transcription from RNA polymerase II promoter
          negative regulation of production of miRNAs involved in gene silencing by miRNA
          regulation of fatty acid transport
          RNA polymerase II regulatory region DNA binding
          RNA polymerase II activating transcription factor binding
          chromatin binding
          transcription corepressor activity
          protein binding
          ligand-dependent nuclear receptor binding
          protein domain specific binding
          estrogen receptor binding
          nuclear hormone receptor binding
          histone deacetylase binding
          peroxisome proliferator activated receptor binding
          transcription regulatory region DNA binding
          retinoid X receptor binding
          thyroid hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0