Search
Search

HPSE| Gene Symbol: HPSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098540.2 | NP_001092010.1 | 10 | 1373 | ||
| NM_001166498.2 | NP_001159970.1 | 9 | 1189 | |||
| NM_001199830.1 | NP_001186759.1 | 9 | 1131 | |||
| NM_006665.5 | NP_006656.2 | 11 | 1411 | |||
| GenBank mRNA | AF084467.1 | - | 10 | 1280 | ||
| AF144325.1 | - | 10 | 1373 | |||
| AF152376.1 | - | 10 | 1288 | |||
| AF155510.1 | - | 11 | 1410 | |||
| AF165154.1 | - | 10 | 1325 | |||
| AK075400.1 | - | 10 | 1337 | |||
| AK222986.1 | - | 10 | 1339 | |||
| AM419200.1 | - | 9 | 1100 | |||
| AY948074.1 | - | 10 | 1279 | |||
| BC051321.1 | - | 11 | 1411 | 6250265 | ||
| FJ517659.1 | - | 10 | 1242 | |||
| FJ517660.1 | - | 9 | 1068 | |||
| GQ337901.1 | - | 9 | 1189 | |||
| GQ337902.1 | - | 10 | 1296 | |||