Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Plcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053456.1 | NP_445908.1 | 5 | 3563 | ||
XM_008767141.1 | XP_008765363.1 | 4 | 7344 | |||
GenBank mRNA | D45920.1 | - | 5 | 3563 |