Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPE SIRNA Assays ›
Gene Symbol:
RPE
Gene Name:
ribulose-5-phosphate-3-epimerase
Gene Aliases:
RPE2-1
Chromosome Location:
Chr.2: 210002565 - 210029156 on Build GRCh38
UniGene ID:
Hs.282260
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224554
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RPE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001278282.1
NP_001265211.1
6
653
NM_001278283.1
NP_001265212.1
7
733
NM_001278286.1
NP_001265215.1
5
577
NM_001278288.1
NP_001265217.1
4
568
NM_001278289.1
NP_001265218.1
5
648
NM_001318926.1
NP_001305855.1
6
702
NM_001318927.1
NP_001305856.1
5
648
NM_001318928.1
NP_001305857.1
6
597
NM_001318929.1
NP_001305858.1
5
648
NM_001318930.1
NP_001305859.1
5
637
NM_001318931.1
NP_001305860.1
5
599
NM_006916.2
NP_008847.1
5
622
NM_199229.2
NP_954699.1
5
648
XM_006712677.3
XP_006712740.1
6
595
GenBank mRNA
AJ224326.1
-
4
390
AK056028.1
-
5
559
AK093658.1
-
7
664
AK096199.1
-
6
484
AK291035.1
-
5
575
AK308741.1
-
5
486
AL833120.1
-
5
571
BC005148.2
-
5
556
3505010
BC016764.1
-
5
585
4107315
BC072401.1
-
5
557
6047438
BG546879.1
-
5
585
BX403091.2
-
6
608
BX643719.1
-
5
574
CN372165.1
-
5
571
CR992340.1
-
5
548
DA237285.1
-
4
495
DA251418.1
-
1
559
DA336224.1
-
5
508
DA592494.1
-
6
525
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
pentose-phosphate shunt
pentose-phosphate shunt, non-oxidative branch
pentose catabolic process
cellular carbohydrate metabolic process
ribulose-phosphate 3-epimerase activity
identical protein binding
protein homodimerization activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline