Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAD SIRNA Assays ›
          Gene Symbol:
          CAD
          Gene Name:
          carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
          Gene Aliases:
          CDG1Z
          Chromosome Location:
          Chr.2: 27217257 - 27243792 on Build GRCh38
          UniGene ID:
          Hs.377010
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s224811
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306079.1 NP_001293008.1 25 4265
          NM_004341.4 NP_004332.2 26 4454
          XM_005264555.3 XP_005264612.1 26 4587
          XM_006712101.2 XP_006712164.1 25 4398
          GenBank mRNA AK226105.1 - 25 4247
          BC014178.2 - 1 171
          BC065510.1 - 26 4454 5551082
          BM927624.1 - 1 194
          BU541191.1 - 5 718
          D78586.1 - 26 4318

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00983188_m1

          Panther Classification:

          Molecular Function -

          hydrolase ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          urea cycle
          liver development
          'de novo' pyrimidine nucleobase biosynthetic process
          UTP biosynthetic process
          arginine biosynthetic process
          glutamine metabolic process
          heart development
          female pregnancy
          lactation
          response to amine
          drug metabolic process
          peptidyl-threonine phosphorylation
          response to caffeine
          organ regeneration
          response to testosterone
          cellular response to drug
          'de novo' UMP biosynthetic process
          pyrimidine nucleoside biosynthetic process
          protein autophosphorylation
          response to cortisol
          cellular response to epidermal growth factor stimulus
          aspartate carbamoyltransferase activity
          carbamoyl-phosphate synthase (ammonia) activity
          carbamoyl-phosphate synthase (glutamine-hydrolyzing) activity
          dihydroorotase activity
          protein kinase activity
          ATP binding
          zinc ion binding
          enzyme binding
          identical protein binding
          aspartate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0