Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164673.1 | NP_001158145.1 | 7 | 875 | ||
NM_001256896.1 | NP_001243825.1 | 4 | 584 | |||
NM_001301071.1 | NP_001288000.1 | 7 | 886 | |||
NM_173660.4 | NP_775931.3 | 7 | 886 | |||
XM_011513435.2 | XP_011511737.1 | 7 | 886 | |||
XM_011513437.2 | XP_011511739.1 | 7 | 875 | |||
XM_017008038.1 | XP_016863527.1 | 5 | 449 | |||
GenBank mRNA | AB220918.1 | - | 7 | 816 | ||
AK091037.1 | - | 7 | 870 | |||
AK096456.1 | - | 4 | 584 | |||
BC043568.1 | - | 3 | 223 | |||
BC062369.1 | - | 3 | 278 | |||
BC131544.1 | - | 7 | 823 | 40085498 | ||
BC141852.1 | - | 7 | 857 | 8860352 |