Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LDHA SIRNA Assays ›
          Gene Symbol:
          LDHA
          Gene Name:
          lactate dehydrogenase A
          Gene Aliases:
          GSD11, HEL-S-133P, LDHM, PIG19
          Chromosome Location:
          Chr.11: 18394389 - 18408218 on Build GRCh38
          UniGene ID:
          Hs.2795
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s229664

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LDHA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135239.1 NP_001128711.1 2, 3 393
          NM_001165414.1 NP_001158886.1 2, 3 490
          NM_001165415.1 NP_001158887.1 2, 3 393
          NM_001165416.1 NP_001158888.1 2, 3 393
          NM_005566.3 NP_005557.1 2, 3 393
          GenBank mRNA AK026515.1 - 2, 3 219
          AK130035.1 - 2, 3 222
          AK130105.1 - 2, 3 217
          AK130587.1 - 2, 3 220
          AK223078.1 - 2, 3 219
          AK296089.1 - 2, 3 219
          AK296667.1 - 2, 3 220
          AK298834.1 - 2, 3 344
          AY423727.1 - 1, 2 152
          BC067223.1 - 2, 3 223 4096518
          BG424022.1 - 1, 2 23
          BG760757.1 - 2, 3 363
          CR456775.1 - 1, 2 121
          CR541714.1 - 1, 2 121
          DA609380.1 - 2, 3 351
          DB502157.1 - 2, 3 394
          DC392310.1 - 2, 3 490
          DC396757.1 - 2, 3 452
          EU794632.1 - 2, 3 218
          X02152.1 - 2, 3 218

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017025, XM_006229232, BC084698, BC168737, BC060587, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001136069, NM_010699, AU079257, CN704214, AK205566, and others.

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          lactate metabolic process
          pyruvate metabolic process
          glycolytic process
          response to nutrient
          response to glucose
          NAD metabolic process
          response to drug
          response to hydrogen peroxide
          positive regulation of apoptotic process
          response to estrogen
          post-embryonic organ development
          response to cAMP
          oxidation-reduction process
          cell-cell adhesion
          L-lactate dehydrogenase activity
          protein binding
          kinase binding
          identical protein binding
          NAD binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0