Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TBC1D3I SIRNA Assays ›
          Gene Symbol:
          TBC1D3I LOC101060389 TBC1D3 TBC1D3B TBC1D3C TBC1D3E TBC1D3F TBC1D3G TBC1D3H TBC1D3K TBC1D3L
          Gene Name:
          TBC1 domain family member 3I
          TBC1 domain family member-like
          TBC1 domain family member 3
          TBC1 domain family member 3B
          TBC1 domain family member 3C
          TBC1 domain family member 3E
          TBC1 domain family member 3F
          TBC1 domain family member 3G
          TBC1 domain family member 3H
          TBC1 domain family member 3K
          TBC1 domain family member 3L
          Gene Aliases:
          -
          TBC1D3D
          PRC17, TBC1D3A, TBC1D3F
          PRC17, TBC1D3I, TBC1D3L
          TBC1D3D
          TBC1DE3
          TBC1D3
          -
          -
          -
          -
          Chromosome Location:
          Chr.17: 36253639 - 36264601 on Build GRCh38
          Chromosome Location:
          Chr.17: 38004033 - 38014908 on Build GRCh38
          Chromosome Location:
          Chr.17: 38181476 - 38192555 on Build GRCh38
          Chromosome Location:
          Chr.17: 36165681 - 36176624 on Build GRCh38
          Chromosome Location:
          Chr.17: 38057693 - 38068634 on Build GRCh38
          Chromosome Location:
          Chr.17: 38119293 - 38138868 on Build GRCh38
          Chromosome Location:
          Chr.17: 36428612 - 36439523 on Build GRCh38
          Chromosome Location:
          Chr.17: 36322242 - 36334759 on Build GRCh38
          Chromosome Location:
          Chr.17: 36377475 - 36388448 on Build GRCh38
          Chromosome Location:
          Chr.17: 37923805 - 37935371 on Build GRCh38
          Chromosome Location:
          Chr.17: 37978155 - 37990691 on Build GRCh38
          UniGene ID:
          Hs.454716
          Hs.454716
          Hs.454716
          Hs.105891
          Hs.735975
          Hs.454716
          Hs.617010
          Hs.454716
          Hs.105891
          Hs.454716
          Hs.454716
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s230583

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TBC1D3I
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291463.1 NP_001278392.1 5 358
          XM_006722223.3 XP_006722286.1 5 629
          XM_006722224.3 XP_006722287.1 4 1874
          XM_006722226.3 XP_006722289.1 4 1874
          XM_011524183.2 XP_011522485.1 4 2057
          XM_011524184.2 XP_011522486.1 4 2057
          XM_011524185.2 XP_011522487.1 4 2057
          GenBank mRNA BC078140.1 - 5 359 5742850
          Gene Symbol: LOC101060389
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291465.1 NP_001278394.1 5 358
          XM_006722248.2 XP_006722311.1 4 1864
          XM_006722249.2 XP_006722312.1 4 1864
          XM_006722250.2 XP_006722313.1 4 1864
          XM_006722252.3 XP_006722315.1 4 1681
          XM_006722253.2 XP_006722316.1 4 1864
          XM_011524176.2 XP_011522478.1 5 471
          XM_017024010.1 XP_016879499.1 4 1864
          GenBank mRNA BI088323.1 - 3 121
          Gene Symbol: TBC1D3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123391.3 NP_001116863.3 5 370
          GenBank mRNA AB449884.1 - 4 258
          AK302064.1 - 6 596
          AK303259.1 - 4 390
          AX775941.1 - 7 619
          AX775943.1 - 7 619
          Gene Symbol: TBC1D3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001417.6 NP_001001417.6 5 404
          XM_005257980.4 XP_005258037.2 5 534
          GenBank mRNA AF540953.1 - 4 258
          AK292257.1 - 5 375
          Gene Symbol: TBC1D3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001418.6 NP_001001418.5 5 398
          XM_006721904.2 XP_006721967.1 4 1864
          XM_006721905.2 XP_006721968.1 4 1864
          XM_006721907.3 XP_006721970.1 4 1681
          XM_006721908.2 XP_006721971.1 4 1864
          XM_011524814.2 XP_011523116.1 5 471
          XM_017024646.1 XP_016880135.1 4 1864
          GenBank mRNA AK313329.1 - 5 398
          Gene Symbol: TBC1D3E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291466.1 NP_001278395.1 5 356
          XM_006722254.1 XP_006722317.1 8 705
          GenBank mRNA AL136860.1 - 5 356
          BC047739.1 - 6 569
          Gene Symbol: TBC1D3F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032258.4 NP_115634.3 5 372
          GenBank mRNA AK225985.1 - 5 363
          AK292344.1 - 5 415
          BC075809.1 - 5 404 5170064
          Gene Symbol: TBC1D3G
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291462.1 NP_001278391.1 5 358
          XM_005276914.3 XP_005276971.1 5 2183
          GenBank mRNA AK292308.1 - 5 408
          AK301823.1 - 5 357
          BC071680.1 - 5 358 5165385
          Gene Symbol: TBC1D3H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123392.3 NP_001116864.3 5 372
          XM_006722055.1 XP_006722118.1 4 1012
          XM_006722056.1 XP_006722119.1 4 1012
          XM_011525172.1 XP_011523474.1 4 1195
          XM_011525173.2 XP_011523475.1 5 566
          XM_011525174.1 XP_011523476.1 4 1195
          XM_011525175.1 XP_011523477.1 4 1195
          XM_011525176.1 XP_011523478.1 4 1195
          GenBank mRNA CB131287.1 - 4 208
          DC398423.1 - 5 373
          Gene Symbol: TBC1D3K
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291464.1 NP_001278393.1 5 358
          XM_006722235.2 XP_006722298.1 4 2636
          XM_006722236.1 XP_006722299.1 5 1208
          XM_006722237.2 XP_006722300.1 4 2453
          XM_006722239.2 XP_006722302.1 4 2453
          XM_011524172.1 XP_011522474.1 4 2636
          XM_011524173.1 XP_011522475.1 4 2636
          XM_011524174.1 XP_011522476.1 4 2636
          GenBank mRNA DC397859.1 - 5 391
          Gene Symbol: TBC1D3L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291459.1 NP_001278388.1 5 372
          XM_006722242.3 XP_006722305.1 4 3612
          XM_006722243.2 XP_006722306.1 4 1864
          XM_006722244.2 XP_006722307.1 4 1864
          XM_006722246.3 XP_006722309.1 4 1012
          XM_006722247.2 XP_006722310.1 4 1864
          XM_011524175.2 XP_011522477.1 5 471
          GenBank mRNA AK095385.1 - 4 1012
          AK122833.1 - 1 1683
          AX775949.1 - 4 1012
          AX775951.1 - 4 1012

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          regulation of cilium assembly
          intracellular protein transport
          regulation of vesicle fusion
          activation of GTPase activity
          GTPase activator activity
          Rab GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0