Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L3 SIRNA Assays ›
Gene Symbol:
EPB41L3
Gene Name:
erythrocyte membrane protein band 4.1 like 3
Gene Aliases:
4.1B, DAL-1, DAL1
Chromosome Location:
Chr.18: 5392381 - 5630666 on Build GRCh38
UniGene ID:
Hs.213394
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23112
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281533.1
NP_001268462.1
15
1969
NM_001281534.1
NP_001268463.1
14
2114
NM_001281535.1
NP_001268464.1
16
1915
NM_012307.3
NP_036439.2
16
2657
XM_011525623.2
XP_011523925.1
15
2150
XM_011525626.2
XP_011523928.1
13
1716
XM_011525628.2
XP_011523930.1
15
2150
XM_011525632.2
XP_011523934.1
14
1752
XM_011525634.2
XP_011523936.1
14
1752
XM_011525637.2
XP_011523939.1
12
1662
XM_017025618.1
XP_016881107.1
13
1716
XM_017025619.1
XP_016881108.1
13
1716
XM_017025620.1
XP_016881109.1
13
1716
XM_017025621.1
XP_016881110.1
12
1662
XM_017025622.1
XP_016881111.1
15
2096
XM_017025623.1
XP_016881112.1
13
1716
XM_017025624.1
XP_016881113.1
14
1752
XM_017025625.1
XP_016881114.1
15
1918
XM_017025626.1
XP_016881115.1
13
1716
XM_017025627.1
XP_016881116.1
15
2150
XM_017025628.1
XP_016881117.1
14
2114
XM_017025629.1
XP_016881118.1
15
2096
XM_017025630.1
XP_016881119.1
15
1864
XM_017025631.1
XP_016881120.1
12
1662
XM_017025632.1
XP_016881121.1
15
2150
XM_017025633.1
XP_016881122.1
15
1918
XM_017025634.1
XP_016881123.1
14
1752
XM_017025635.1
XP_016881124.1
13
1716
XM_017025636.1
XP_016881125.1
15
2150
XM_017025637.1
XP_016881126.1
15
2150
XM_017025638.1
XP_016881127.1
13
1716
XM_017025639.1
XP_016881128.1
15
2096
XM_017025640.1
XP_016881129.1
14
2114
XM_017025641.1
XP_016881130.1
12
1662
XM_017025642.1
XP_016881131.1
15
2096
XM_017025643.1
XP_016881132.1
15
2150
XM_017025644.1
XP_016881133.1
14
2060
XM_017025645.1
XP_016881134.1
15
2009
XM_017025646.1
XP_016881135.1
14
2114
XM_017025647.1
XP_016881136.1
14
1973
XM_017025648.1
XP_016881137.1
15
2009
XM_017025649.1
XP_016881138.1
15
1955
XM_017025650.1
XP_016881139.1
15
2009
XM_017025651.1
XP_016881140.1
14
1973
XM_017025652.1
XP_016881141.1
15
1955
XM_017025653.1
XP_016881142.1
15
1955
XM_017025654.1
XP_016881143.1
15
2009
XM_017025655.1
XP_016881144.1
14
1919
XM_017025656.1
XP_016881145.1
15
2009
XM_017025657.1
XP_016881146.1
15
2009
XM_017025658.1
XP_016881147.1
15
1955
XM_017025659.1
XP_016881148.1
14
1973
XM_017025660.1
XP_016881149.1
15
1955
GenBank mRNA
AB023204.1
-
16
2334
AF069072.1
-
14
1771
AK094952.1
-
4
248
AK128531.1
-
16
1782
AK292583.1
-
17
1915
AK294610.1
-
4
242
AK294690.1
-
3
239
AK295453.1
-
4
236
AK297406.1
-
15
1969
AK303679.1
-
5
432
AK307751.1
-
5
424
AK314438.1
-
13
1441
BC006141.1
-
15
2085
3936453
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
protein localization to paranode region of axon
apoptotic process
cytoskeletal anchoring at plasma membrane
biological_process
regulation of cell shape
cortical cytoskeleton organization
cortical actin cytoskeleton organization
paranodal junction assembly
myelin maintenance
neuron projection morphogenesis
protein localization to juxtaparanode region of axon
protein localization to plasma membrane
actin binding
structural constituent of cytoskeleton
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0