Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GAPDH SIRNA Assays ›
          Gene Symbol:
          Gapdh Gm10481 Gm12070 Gm5069 Gm5523 Gm6981
          Gene Name:
          glyceraldehyde-3-phosphate dehydrogenase
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          Gene Aliases:
          Gapd
          ENSMUSG00000073212
          OTTMUSG00000005300
          EG277333
          EG433273
          EG629557
          Chromosome Location:
          Chr.6: 125161338 - 125166511 on Build GRCm38
          Chromosome Location:
          Chr.5: 13042242 - 13043452 on Build GRCm38
          Chromosome Location:
          Chr.11: 26785108 - 26787860 on Build GRCm38
          Chromosome Location:
          Chr.1: 180326969 - 180330549 on Build GRCm38
          Chromosome Location:
          Chr.1: 13785685 - 13786929 on Build GRCm38
          Chromosome Location:
          Chr.9: 52002061 - 52048705 on Build GRCm38
          UniGene ID:
          Mm.304088
          Mm.309092
          Mm.347777
          Mm.389223
          Mm.304088
          Mm.422775
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s234319

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gapdh
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289726.1 NP_001276655.1 6 993
          NM_008084.3 NP_032110.1 6 1141
          GenBank mRNA AK002273.1 - 6 974
          AK081405.1 - 6 974
          AK140794.1 - 6 974
          AK144690.1 - 6 995
          AK146435.1 - 6 974
          AK147738.1 - 6 974
          AK147891.1 - 6 974
          AK160399.1 - 6 974
          AK160753.1 - 6 971
          AK164415.1 - 6 972
          AK168217.1 - 6 974
          AK169742.1 - 1 2762
          AK178056.1 - 1 17
          AK190093.1 - 1 17
          AK190330.1 - 1 17
          AK190481.1 - 1 17
          AK190634.1 - 1 18
          AK191255.1 - 1 17
          AK191361.1 - 1 17
          AK191558.1 - 1 17
          AK191930.1 - 1 17
          AK192123.1 - 1 17
          AK192402.1 - 1 17
          AK192522.1 - 1 17
          AK192930.1 - 17
          AK193656.1 - 1 17
          AK194000.1 - 1 17
          AK194274.1 - 17
          AK194309.1 - 1 17
          AK194315.1 - 1 17
          AK194885.1 - 1 17
          AK195724.1 - 1 17
          AK196170.1 - 1 17
          AK196250.1 - 1 17
          AK196299.1 - 1 17
          AK196324.1 - 1 17
          AK196391.1 - 1 17
          AK196504.1 - 17
          AK196631.1 - 1 17
          AK197155.1 - 1 17
          AK197552.1 - 1 17
          AK198019.1 - 1 17
          AK198344.1 - 1 17
          AK198622.1 - 1 17
          AK198692.1 - 1 17
          AK199039.1 - 1 17
          AK199353.1 - 1 17
          AK200221.1 - 1 17
          AK200222.1 - 1 17
          AK201249.1 - 1 17
          AK201401.1 - 1 17
          AK201876.1 - 1 17
          AK202784.1 - 1 17
          AK203035.1 - 1 17
          AK203145.1 - 1 18
          AK203274.1 - 1 17
          AK203291.1 - 1 16
          AK203511.1 - 1 17
          AK203514.1 - 1 17
          AK203950.1 - 1 17
          AK204125.1 - 1 17
          AK204204.1 - 1 17
          AK204369.1 - 1 17
          AK204431.1 - 17
          AK204435.1 - 1 17
          AK204477.1 - 1 17
          AK204659.1 - 1 17
          AK205615.1 - 1 17
          AK206172.1 - 1 17
          AK214525.1 - 1 17
          AK218826.1 - 17
          BC020407.1 - 6 962
          BC023196.1 - 6 972
          BC082592.1 - 6 951 30634381
          BC083065.1 - 6 950 5371133
          BC083079.1 - 6 956 6493407
          BC083080.1 - 6 958 6494145
          BC083149.1 - 937 30459585
          BC085274.1 - 959 6530979
          BC085275.1 - 6 968 30444086
          BC091736.1 - 496
          BC091768.1 - 937 4159824
          BC092252.1 - 6 960 5044783
          BC092264.1 - 962 2655526
          BC092267.1 - 948 4010884
          BC092294.1 - 951 5068049
          BC093508.1 - 931 4975394
          BC094037.1 - 938 5009636
          BC095932.1 - 6 962 30733532
          BC096042.1 - 325
          BC096440.1 - 6 962 2649847
          BC096590.1 - 962 6486174
          BC110311.1 - 6 963 5340298
          BC145810.1 - 6 944 40131113
          BC145812.1 - 6 944 40131115
          BI851476.1 - 3 506
          GU214026.1 - 6 972
          KF982306.1 - 145
          M32599.1 - 6 947
          Gene Symbol: Gm10481
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK160495.1 - 334
          Gene Symbol: Gm12070
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC098095.1 - 2 1320
          Gene Symbol: Gm5069
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC064681.1 - 2 1026
          Gene Symbol: Gm5523
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK190063.1 - 17
          Gene Symbol: Gm6981
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK132984.1 - 3 1152
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AK203228.1 - 17 Chr. - - [-] on Build

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017008, M11561, BC087743, AB017801, BC059110, and others.

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          carbohydrate metabolic process
          gluconeogenesis
          glycolytic process
          apoptotic process
          multicellular organism development
          negative regulation of translation
          peptidyl-cysteine S-trans-nitrosylation
          protein stabilization
          neuron apoptotic process
          oxidation-reduction process
          response to ammonium ion
          cellular response to interferon-gamma
          glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
          microtubule binding
          peptidyl-cysteine S-nitrosylase activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-wksrx:80/100.66.72.56:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0