Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other H2AFX SIRNA Assays ›
          Gene Symbol:
          H2afx H2afj Hist1h2aa Hist1h2ab Hist1h2ac Hist1h2ad Hist1h2ae Hist1h2af Hist1h2ag Hist1h2ah Hist1h2ai Hist1h2ak Hist1h2al Hist1h2an Hist1h2ao Hist1h2ap Hist2h2aa1 Hist2h2aa2 Hist2h2ac Hist3h2a
          Gene Name:
          H2A histone family, member X
          H2A histone family, member J
          histone cluster 1, H2aa
          histone cluster 1, H2ab
          histone cluster 1, H2ac
          histone cluster 1, H2ad
          histone cluster 1, H2ae
          histone cluster 1, H2af
          histone cluster 1, H2ag
          histone cluster 1, H2ah
          histone cluster 1, H2ai
          histone cluster 1, H2ak
          histone cluster 1, H2al
          histone cluster 1, H2an
          histone cluster 1, H2ao
          histone cluster 1, H2ap
          histone cluster 2, H2aa1
          histone cluster 2, H2aa2
          histone cluster 2, H2ac
          histone cluster 3, H2a
          Gene Aliases:
          AW228881, H2A.X, H2ax, Hist5-2ax, gammaH2ax
          E130307C13
          Th2a
          H2A.1, H2a-53
          -
          -
          1190022L06
          H2a-221
          -
          -
          H2a-291A
          -
          EG667728
          -
          -
          ENSMUSG00000060262, Gm11276, OTTMUSG00000000448
          AI448581, H2a-614, H2a-615, Hist2
          H2a-614, H2a-615
          H2a-613b
          -
          Chromosome Location:
          Chr.9: 44334715 - 44336077 on Build GRCm38
          Chromosome Location:
          Chr.6: 136808248 - 136810074 on Build GRCm38
          Chromosome Location:
          Chr.13: 23934462 - 23934913 on Build GRCm38
          Chromosome Location:
          Chr.13: 23751088 - 23751592 on Build GRCm38
          Chromosome Location:
          Chr.13: 23683473 - 23683959 on Build GRCm38
          Chromosome Location:
          Chr.13: 23574381 - 23574915 on Build GRCm38
          Chromosome Location:
          Chr.13: 23570663 - 23571220 on Build GRCm38
          Chromosome Location:
          Chr.13: 23533911 - 23534378 on Build GRCm38
          Chromosome Location:
          Chr.13: 22042478 - 22042949 on Build GRCm38
          Chromosome Location:
          Chr.13: 22035122 - 22035643 on Build GRCm38
          Chromosome Location:
          Chr.13: 21716412 - 21716859 on Build GRCm38
          Chromosome Location:
          Chr.13: 21753377 - 21753912 on Build GRCm38
          Chromosome Location:
          Chr.13: 51202636 - 51203068 on Build GRCm38
          Chromosome Location:
          Chr.13: 21786772 - 21787218 on Build GRCm38
          Chromosome Location:
          Chr.13: 21810367 - 21810918 on Build GRCm38
          Chromosome Location:
          Chr.13: 21833024 - 21833475 on Build GRCm38
          Chromosome Location:
          Chr.3: 96245497 - 96246052 on Build GRCm38
          Chromosome Location:
          Chr.3: 96239779 - 96240374 on Build GRCm38
          Chromosome Location:
          Chr.3: 96220413 - 96220880 on Build GRCm38
          Chromosome Location:
          Chr.11: 58954685 - 58955192 on Build GRCm38
          UniGene ID:
          Mm.245931
          Mm.154564
          Mm.377876
          Mm.423402
          Mm.261973
          Mm.390553
          Mm.261665
          Mm.377878
          Mm.263165
          Mm.250564
          Mm.14767
          Mm.370338
          -
          -
          Mm.250564
          Mm.250564
          Mm.478832
          Mm.478832
          Mm.358954
          Mm.212549
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234516

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: H2afx
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010436.2 NP_034566.1 1 321
          GenBank mRNA AK008124.1 - 1 341
          AK088040.1 - 1 344
          BC005468.1 - 1 315 3490058
          BC010336.1 - 1 321 3156946
          X58069.1 - 1 325
          Gene Symbol: H2afj
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_177688.4 NP_808356.1 1 366
          GenBank mRNA AK053755.1 - 1 291
          AV498351.1 - 1 365
          BC024397.1 - 1 326 5055276
          BC099606.1 - 1 348 1383389
          Gene Symbol: Hist1h2aa
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175658.2 NP_783589.1 1 274
          GenBank mRNA BC107354.1 - 1 274 40055855
          BC107355.1 - 1 274 40055860
          Gene Symbol: Hist1h2ab
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175660.3 NP_783591.2 1 319
          GenBank mRNA BC117110.1 - 1 318 40126429
          Gene Symbol: Hist1h2ac
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178189.5 NP_835496.1 1 316
          GenBank mRNA AK033518.1 - 318
          Gene Symbol: Hist1h2ad
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178188.4 NP_835495.1 1 364
          GenBank mRNA BC119295.1 - 1 364 8734044
          BC119297.1 - 1 364 8734046
          Gene Symbol: Hist1h2ae
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178187.4 NP_835494.1 1 386
          GenBank mRNA AK028026.1 - 1 387
          BC058544.1 - 1 305 5711765
          BC076498.1 - 1 304 5713252
          Gene Symbol: Hist1h2af
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175661.2 NP_783592.1 1 274
          GenBank mRNA BC125011.1 - 1 274 40051842
          Gene Symbol: Hist1h2ag
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178186.3 NP_835493.1 1 296
          GenBank mRNA BC065803.1 - 1 296 1263745
          Gene Symbol: Hist1h2ah
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175659.2 NP_783590.1 1 365
          GenBank mRNA BC125248.1 - 1 273
          BC134370.1 - 1 274 40100857
          BC139479.1 - 1 365 8862501
          BC146008.1 - 1 365 40131311
          Gene Symbol: Hist1h2ai
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178182.2 NP_835489.1 1 284
          GenBank mRNA BC099406.1 - 284 30789778
          BC116373.2 - 1 274 40100893
          BC127164.1 - 1 274 40100895
          Gene Symbol: Hist1h2ak
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178183.2 NP_835490.1 1 359
          GenBank mRNA BC119335.1 - 1 359 8734084
          Gene Symbol: Hist1h2al
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK077055.1 - 132
          Gene Symbol: Hist1h2an
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178184.2 NP_835491.1 1 274
          Gene Symbol: Hist1h2ao
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177544.2 NP_001171015.1 1 374
          Gene Symbol: Hist1h2ap
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178185.2 NP_835492.1 1 274
          GenBank mRNA AK145443.1 - 1 307
          AK146846.1 - 1 305
          BC062251.1 - 1 286 1224905
          BC090402.1 - 1 286 5150365
          BC110626.1 - 1 274 40051841
          BC133661.1 - 1 274 40051853
          BC147161.1 - 1 374 9056189
          BC147162.1 - 1 374 9056203
          Gene Symbol: Hist2h2aa1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013549.2 NP_038577.1 1 358
          GenBank mRNA AK002725.1 - 1 318
          AK006728.1 - 1 317
          BC010564.1 - 1 301 3582122
          BC053966.1 - 1 310
          BC062255.1 - 1 301 1448126
          BC064002.1 - 300
          BC089519.1 - 1 358 6771160
          BC147778.1 - 1 355 9087498
          BC147781.1 - 1 355 9087501
          Z30940.1 - 1 296
          Gene Symbol: Hist2h2aa2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178212.3 NP_835584.1 1 398
          Gene Symbol: Hist2h2ac
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175662.2 NP_783593.1 1 310
          GenBank mRNA BC117094.1 - 1 310 40126413
          BC117096.1 - 1 310 40126415
          Gene Symbol: Hist3h2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178218.4 NP_835736.1 1 327
          GenBank mRNA AK051448.1 - 328
          AK077568.1 - 1 328
          AK082083.1 - 331
          AK087537.1 - 329
          BC058119.1 - 300 6819179
          BC063781.1 - 294 6493838

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_033445, NM_003512, NM_003511, NM_080596, NM_002105, and others.
          This siRNA also targets the following transcripts in Rat: NM_001109423, NM_021840, NM_001109291, NM_021839, XM_003751711, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          double-strand break repair via homologous recombination
          DNA repair
          DNA recombination
          cellular response to DNA damage stimulus
          cell cycle
          spermatogenesis
          meiotic cell cycle
          cellular response to gamma radiation
          biological_process
          chromatin organization
          negative regulation of cell proliferation
          nucleosome disassembly
          UV-damage excision repair
          DNA binding
          damaged DNA binding
          protein binding
          enzyme binding
          histone binding
          protein heterodimerization activity
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0