Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADM1 SIRNA Assays ›
Gene Symbol:
CADM1
Gene Name:
cell adhesion molecule 1
Gene Aliases:
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
Chromosome Location:
Chr.11: 115173625 - 115504523 on Build GRCh38
UniGene ID:
Hs.370510
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24326
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098517.1
NP_001091987.1
7
1109
NM_001301043.1
NP_001287972.1
7
1109
NM_001301044.1
NP_001287973.1
7
1109
NM_001301045.1
NP_001287974.1
7
1109
NM_014333.3
NP_055148.3
7
1109
XM_005271494.2
XP_005271551.1
7
1002
XM_017017457.1
XP_016872946.1
7
1068
XM_017017458.1
XP_016872947.1
7
875
XM_017017459.1
XP_016872948.1
8
3062
XM_017017460.1
XP_016872949.1
8
3062
XM_017017461.1
XP_016872950.1
8
1076
GenBank mRNA
AB094146.1
-
7
980
AF132811.1
-
7
983
AK075502.1
-
7
999
AK315245.1
-
7
1042
AY309502.1
-
2
177
AY358334.1
-
7
995
BC035930.1
-
7
1001
BC047021.1
-
7
1109
5248100
BC125102.1
-
7
984
BC125103.1
-
7
983
40118330
CR457157.1
-
7
980
HE586496.1
-
7
983
HE586497.1
-
7
983
HE586498.1
-
7
983
HE586499.1
-
7
983
HE586500.1
-
8
1069
HE586501.1
-
1
100
HE586503.1
-
8
1011
KJ534791.1
-
7
980
KJ534792.1
-
7
980
KJ534793.1
-
7
980
KJ534794.1
-
7
980
KJ534795.1
-
7
980
KJ534796.1
-
7
980
KJ534797.1
-
7
980
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
T cell mediated cytotoxicity
apoptotic process
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
multicellular organism development
spermatogenesis
cell recognition
cell differentiation
adherens junction organization
susceptibility to natural killer cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of cytokine secretion
activated T cell proliferation
detection of stimulus
receptor activity
receptor binding
protein binding
PDZ domain binding
protein homodimerization activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0