Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF302 SIRNA Assays ›
          Gene Symbol:
          ZNF302
          Gene Name:
          zinc finger protein 302
          Gene Aliases:
          HSD16, MST154, MSTP154, ZNF135L, ZNF140L, ZNF327
          Chromosome Location:
          Chr.19: 34677619 - 34686397 on Build GRCh38
          UniGene ID:
          Hs.436350
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s31740

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF302
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012320.2 NP_001012320.1 5 558
          NM_001289181.1 NP_001276110.1 6 713
          NM_001289182.1 NP_001276111.1 5 831
          NM_001289183.1 NP_001276112.1 5 521
          NM_001289184.1 NP_001276113.1 5 584
          NM_001289185.1 NP_001276114.1 5 561
          NM_001289186.1 NP_001276115.1 5 479
          NM_001289187.1 NP_001276116.1 5 828
          NM_001289188.1 NP_001276117.1 5 850
          NM_001289189.1 NP_001276118.1 5 844
          NM_001289190.1 NP_001276119.1 5 781
          NM_001289191.1 NP_001276120.1 5 818
          NM_001289192.1 NP_001276121.1 5 841
          NM_018443.3 NP_060913.2 5 581
          NM_018675.2 NP_061145.2 5 1088
          NR_110322.1 3 376
          XM_011527111.2 XP_011525413.1 5 841
          XM_017026978.1 XP_016882467.1 6 954
          XM_017026979.1 XP_016882468.1 6 951
          XM_017026980.1 XP_016882469.1 6 645
          XM_017026981.1 XP_016882470.1 6 608
          XM_017026982.1 XP_016882471.1 5 456
          XM_017026986.1 XP_016882475.1 5 1082
          GenBank mRNA AF155656.1 - 5 533
          AF190157.1 - 5 520
          AF265236.1 - 6 868
          AF326206.1 - 5 631
          AK074901.1 - 5 531
          AK122855.1 - 3 1780
          AK131278.1 - 6 635
          AK297551.1 - 5 764
          AK315241.1 - 5 523
          AL515741.3 - 5 535
          AL834318.1 - 5 535
          AY251166.1 - 4 326
          BC024176.2 - 5 633 4822400
          DA297006.1 - 5 531
          DA749741.1 - 5 482
          DA773435.1 - 3 370
          DA780378.1 - 5 440
          DA792042.1 - 5 407
          DN997090.1 - 5 575

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0