Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCBP4 SIRNA Assays ›
Gene Symbol:
PCBP4
Gene Name:
poly(rC) binding protein 4
Gene Aliases:
CBP, LIP4, MCG10
Chromosome Location:
Chr.3: 51957454 - 51967466 on Build GRCh38
UniGene ID:
Hs.20930
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32630
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCBP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174100.1
NP_001167571.1
4, 5
469
NM_020418.3
NP_065151.2
4, 5
372
NM_033008.2
NP_127501.1
3, 4
321
NM_033010.2
NP_127503.1
3, 4
263
XM_005265330.2
XP_005265387.1
4, 5
2276
XM_005265331.2
XP_005265388.1
4, 5
395
XM_005265332.2
XP_005265389.1
4, 5
2276
XM_006713268.2
XP_006713331.1
4, 5
2084
XM_006713269.1
XP_006713332.1
5, 6
540
XM_006713270.1
XP_006713333.1
3, 4
312
XM_006713271.2
XP_006713334.1
4, 5
2276
XM_017006916.1
XP_016862405.1
5, 6
697
XM_017006917.1
XP_016862406.1
4, 5
2084
XM_017006918.1
XP_016862407.1
3, 4
1936
XM_017006919.1
XP_016862408.1
3, 4
312
XM_017006920.1
XP_016862409.1
3, 4
312
GenBank mRNA
AF092441.1
-
3, 4
235
AF176330.1
-
3, 4
263
AF257770.1
-
2, 3
512
AF257771.1
-
2, 3
512
AK001244.1
-
4, 5
644
AK023993.1
-
3, 4
310
AK225815.1
-
4, 5
448
BC003008.1
-
3, 4
284
2822683
BC004153.2
-
3, 4
284
2822683
BC017098.1
-
4, 5
367
3508303
BX647811.1
-
3, 4
627
DA701799.1
-
5, 6
439
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
regulation of mRNA stability
DNA binding
RNA binding
mRNA 3'-UTR binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0