Search
Search

CPT1A| Gene Symbol: CPT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031847.2 | NP_001027017.1 | 16 | 2154 | ||
| NM_001876.3 | NP_001867.2 | 16 | 2154 | |||
| XM_005273762.2 | XP_005273819.1 | 16 | 2130 | |||
| XM_005273763.1 | XP_005273820.1 | 16 | 2130 | |||
| XM_017017220.1 | XP_016872709.1 | 16 | 2263 | |||
| GenBank mRNA | AK025307.1 | - | 11 | 1440 | ||
| AK172798.1 | - | 16 | 2142 | |||
| AK314301.1 | - | 16 | 2093 | |||
| BC000185.1 | - | 16 | 2029 | 3352642 | ||
| BT009791.1 | - | 15 | 1984 | |||
| L39211.1 | - | 16 | 2060 | |||