Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152343.2 | NP_689556.2 | 5 | 1204 | ||
GenBank mRNA | AF433663.1 | - | 5 | 1170 | ||
AK058143.1 | - | 5 | 1204 | |||
AK302184.1 | - | 7 | 1827 | |||
BC028124.1 | - | 5 | 1159 | 5166606 |