Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DRD1 SIRNA Assays ›
Gene Symbol:
DRD1
Gene Name:
dopamine receptor D1
Gene Aliases:
DADR, DRD1A
Chromosome Location:
Chr.5: 175440672 - 175444160 on Build GRCh38
UniGene ID:
Hs.2624
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4282
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DRD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000794.3
NP_000785.1
2
2209
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
conditioned taste aversion
behavioral fear response
synaptic transmission, dopaminergic
response to amphetamine
negative regulation of protein kinase activity
protein import into nucleus
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-activating dopamine receptor signaling pathway
synapse assembly
learning
memory
mating behavior
grooming behavior
adult walking behavior
visual learning
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
astrocyte development
response to activity
dopamine transport
transmission of nerve impulse
neuronal action potential
regulation of vasoconstriction
calcium-mediated signaling
dentate gyrus development
striatum development
orbitofrontal cortex development
cerebral cortex GABAergic interneuron migration
positive regulation of cell migration
negative regulation of cell migration
peristalsis
positive regulation of cAMP biosynthetic process
response to food
response to estradiol
response to retinoic acid
cellular response to insulin stimulus
response to nicotine
operant conditioning
social behavior
response to antidepressant
regulation of dopamine metabolic process
vasodilation
negative regulation of circadian sleep/wake cycle, sleep
dopamine metabolic process
response to drug
maternal behavior
response to amino acid
positive regulation of potassium ion transport
response to morphine
response to ethanol
positive regulation of membrane potential
glucose import
habituation
sensitization
behavioral response to cocaine
regulation of long-term neuronal synaptic plasticity
response to steroid hormone
positive regulation of release of sequestered calcium ion into cytosol
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
regulation of dopamine uptake involved in synaptic transmission
positive regulation of synaptic transmission, glutamatergic
prepulse inhibition
phospholipase C-activating dopamine receptor signaling pathway
long-term synaptic potentiation
long term synaptic depression
cellular response to hypoxia
cellular response to catecholamine stimulus
positive regulation of long-term synaptic potentiation
positive regulation of feeding behavior
dopamine neurotransmitter receptor activity, coupled via Gs
G-protein alpha-subunit binding
dopamine neurotransmitter receptor activity
protein binding
drug binding
protein phosphatase binding
angiotensin receptor binding
D3 dopamine receptor binding
protein complex binding
dopamine binding
protein heterodimerization activity
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0