Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGFR2 SIRNA Assays ›
Gene Symbol:
FGFR2
Gene Name:
fibroblast growth factor receptor 2
Gene Aliases:
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
Chromosome Location:
Chr.10: 121478330 - 121598656 on Build GRCh38
UniGene ID:
Hs.533683
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s529189
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGFR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000141.4
NP_000132.3
14
2544
NM_001144913.1
NP_001138385.1
13
2050
NM_001144914.1
NP_001138386.1
11
1711
NM_001144915.1
NP_001138387.1
13
1949
NM_001144916.1
NP_001138388.1
11
1993
NM_001144917.1
NP_001138389.1
12
2196
NM_001144918.1
NP_001138390.1
12
2193
NM_001144919.1
NP_001138391.1
13
2280
NM_001320654.1
NP_001307583.1
13
1928
NM_001320658.1
NP_001307587.1
14
2538
NM_022970.3
NP_075259.4
14
2547
NM_023029.2
NP_075418.1
12
1780
NR_073009.1
13
2347
XM_006717708.2
XP_006717771.1
14
2739
XM_006717710.3
XP_006717773.1
14
2745
XM_006717711.2
XP_006717774.1
12
1783
XM_006717712.2
XP_006717775.1
11
1705
XM_017015920.1
XP_016871409.1
14
2739
XM_017015921.1
XP_016871410.1
14
2736
XM_017015922.1
XP_016871411.1
12
1777
XM_017015923.1
XP_016871412.1
12
1774
XM_017015924.1
XP_016871413.1
12
2397
XM_017015925.1
XP_016871414.1
12
2391
GenBank mRNA
AB030073.1
-
14
2318
AB030074.1
-
14
2318
AB030075.1
-
14
2318
AB030076.1
-
14
2318
AB030077.1
-
14
2318
AB030078.1
-
14
2318
AB084153.1
-
11
1561
AK026508.1
-
13
2254
AK294026.1
-
14
1949
AK308621.1
-
11
1993
BC037338.2
-
13
1928
BC039243.1
-
12
2193
5277896
EF057068.1
-
12
1630
KP192323.1
-
12
1711
KP192324.1
-
12
1717
KP192325.1
-
9
1342
KP192326.1
-
12
1708
KP192327.1
-
12
1714
KP192328.1
-
13
1924
KP192329.1
-
11
1579
KP192330.1
-
12
1657
KP192331.1
-
13
1930
M55614.1
-
13
1912
M80634.1
-
14
2318
M87770.1
-
14
2170
M87771.1
-
12
2036
M97193.1
-
14
2511
X52832.1
-
14
2076
X56191.1
-
13
1924
Z71929.1
-
13
1909
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
angiogenesis
ureteric bud development
in utero embryonic development
epithelial to mesenchymal transition
positive regulation of mesenchymal cell proliferation
outflow tract septum morphogenesis
membranous septum morphogenesis
apoptotic process
cell-cell signaling
axonogenesis
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of smoothened signaling pathway
post-embryonic development
embryonic pattern specification
organ morphogenesis
regulation of cell fate commitment
positive regulation of phospholipase activity
regulation of phosphatidylinositol 3-kinase signaling
morphogenesis of embryonic epithelium
peptidyl-tyrosine phosphorylation
orbitofrontal cortex development
ventricular zone neuroblast division
pyramidal neuron development
gland morphogenesis
positive regulation of Wnt signaling pathway
bone mineralization
lung development
epithelial cell differentiation
midbrain development
otic vesicle formation
hair follicle morphogenesis
lacrimal gland development
regulation of osteoblast proliferation
multicellular organism growth
organ growth
fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
fibroblast growth factor receptor signaling pathway involved in hemopoiesis
fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
regulation of multicellular organism growth
regulation of fibroblast growth factor receptor signaling pathway
inner ear morphogenesis
odontogenesis
positive regulation of MAPK cascade
positive regulation of GTPase activity
cell fate commitment
regulation of osteoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
lung alveolus development
mesodermal cell differentiation
embryonic digestive tract morphogenesis
embryonic organ morphogenesis
digestive tract development
embryonic organ development
reproductive structure development
positive regulation of smooth muscle cell proliferation
embryonic cranial skeleton morphogenesis
skeletal system morphogenesis
epidermis morphogenesis
branching morphogenesis of a nerve
mesenchymal cell differentiation
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
regulation of smooth muscle cell differentiation
positive regulation of cell division
ventricular cardiac muscle tissue morphogenesis
positive regulation of cardiac muscle cell proliferation
limb bud formation
bone development
bone morphogenesis
branching involved in prostate gland morphogenesis
branching involved in salivary gland morphogenesis
bud elongation involved in lung branching
lung lobe morphogenesis
lung-associated mesenchyme development
positive regulation of epithelial cell proliferation involved in lung morphogenesis
prostate gland morphogenesis
prostate epithelial cord elongation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
fibroblast growth factor receptor signaling pathway involved in mammary gland specification
lateral sprouting from an epithelium
mammary gland bud formation
epithelial cell proliferation involved in salivary gland morphogenesis
branch elongation involved in salivary gland morphogenesis
branching involved in labyrinthine layer morphogenesis
regulation of branching involved in prostate gland morphogenesis
regulation of morphogenesis of a branching structure
mesenchymal cell differentiation involved in lung development
mesenchymal cell proliferation involved in lung development
regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
positive regulation of canonical Wnt signaling pathway
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lclp9:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0