Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MRE11A SIRNA Assays ›
          Gene Symbol:
          MRE11A
          Gene Name:
          MRE11 homolog A, double strand break repair nuclease
          Gene Aliases:
          ATLD, HNGS1, MRE11, MRE11B
          Chromosome Location:
          Chr.11: 94415570 - 94512701 on Build GRCh38
          UniGene ID:
          Hs.192649
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532541

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MRE11A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005590.3 NP_005581.2 13, 14 1801
          NM_005591.3 NP_005582.1 13, 14 1694
          XM_005274008.3 XP_005274065.1 11, 12 1409
          XM_006718842.3 XP_006718905.1 13, 14 1801
          XM_011542837.2 XP_011541139.1 13, 14 2223
          XM_017017772.1 XP_016873261.1 13, 14 1801
          XM_017017773.1 XP_016873262.1 13, 14 1694
          GenBank mRNA AF022778.1 - 13, 14 1675
          AF073362.1 - 12, 13 1505
          AK095388.1 - 13, 14 1803
          AK308318.1 - 13, 14 1692
          BC017823.1 - 13, 14 1737
          BC063458.1 - 12, 13 1602 5181691
          U37359.1 - 13, 14 1676

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of mitotic recombination
          double-strand break repair via homologous recombination
          DNA double-strand break processing
          DNA synthesis involved in DNA repair
          strand displacement
          DNA replication
          DNA repair
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          DNA recombination
          cellular response to DNA damage stimulus
          telomere maintenance via telomerase
          sister chromatid cohesion
          mitotic G2 DNA damage checkpoint
          synapsis
          reciprocal meiotic recombination
          heart development
          cell proliferation
          viral process
          intra-S DNA damage checkpoint
          telomeric 3' overhang formation
          positive regulation of protein autophosphorylation
          positive regulation of telomere maintenance
          positive regulation of type I interferon production
          DNA duplex unwinding
          negative regulation of DNA endoreduplication
          positive regulation of kinase activity
          negative regulation of apoptotic process
          negative regulation of viral entry into host cell
          nucleic acid phosphodiester bond hydrolysis
          cell-cell adhesion
          regulation of signal transduction by p53 class mediator
          single-stranded DNA endodeoxyribonuclease activity
          DNA binding
          double-stranded DNA binding
          ATP-dependent DNA helicase activity
          nuclease activity
          endodeoxyribonuclease activity
          protein binding
          protein C-terminus binding
          3'-5' exonuclease activity
          manganese ion binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0