Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RELA SIRNA Assays ›
          Gene Symbol:
          RELA
          Gene Name:
          RELA proto-oncogene, NF-kB subunit
          Gene Aliases:
          NFKB3, p65
          Chromosome Location:
          Chr.11: 65653596 - 65662972 on Build GRCh38
          UniGene ID:
          Hs.502875
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s535313

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RELA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145138.1 NP_001138610.1 11 1743
          NM_001243984.1 NP_001230913.1 11 1545
          NM_001243985.1 NP_001230914.1 12 1443
          NM_021975.3 NP_068810.3 11 1752
          XM_011545206.1 XP_011543508.1 10 1520
          XM_011545207.1 XP_011543509.1 9 1415
          GenBank mRNA AK308478.1 - 10 1798
          BC011603.1 - 12 1184
          BC014095.2 - 10 1343
          BC033522.1 - 10 1609 5162908
          BC110830.1 - 10 2362 6019711
          L19067.1 - 10 1608
          M62399.1 - 11 1691

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: BC079457, AY307375.
          This siRNA also targets the following transcripts in Mouse: M61909, AK157076, AK154630, BC094053, AK158072, and others.

          Panther Classification:

          Molecular Function -

          Rel homology transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          liver development
          hair follicle development
          stimulatory C-type lectin receptor signaling pathway
          acetaldehyde metabolic process
          transcription from RNA polymerase II promoter
          inflammatory response
          cellular defense response
          aging
          positive regulation of cell proliferation
          organ morphogenesis
          response to organic substance
          response to UV-B
          positive regulation of Schwann cell differentiation
          cytokine-mediated signaling pathway
          membrane protein intracellular domain proteolysis
          positive regulation of chondrocyte differentiation
          positive regulation of type I interferon production
          response to muramyl dipeptide
          response to progesterone
          response to insulin
          response to cobalamin
          cellular response to hepatocyte growth factor stimulus
          response to muscle stretch
          Fc-epsilon receptor signaling pathway
          negative regulation of protein catabolic process
          response to drug
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to amino acid
          response to morphine
          positive regulation of interleukin-12 biosynthetic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin receptor signaling pathway
          regulation of inflammatory response
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          response to cAMP
          defense response to virus
          cellular response to hydrogen peroxide
          nucleotide-binding oligomerization domain containing 2 signaling pathway
          response to interleukin-1
          cellular response to lipopolysaccharide
          cellular response to nicotine
          cellular response to interleukin-1
          cellular response to interleukin-6
          cellular response to tumor necrosis factor
          cellular response to peptide hormone stimulus
          negative regulation of NIK/NF-kappaB signaling
          positive regulation of miRNA metabolic process
          negative regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          transcription factor binding
          protein kinase binding
          chromatin DNA binding
          ubiquitin protein ligase binding
          protein complex binding
          activating transcription factor binding
          phosphate ion binding
          identical protein binding
          protein homodimerization activity
          actinin binding
          histone deacetylase binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          protein N-terminus binding
          NF-kappaB binding
          repressing transcription factor binding
          ankyrin repeat binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0