Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HFE SIRNA Assays ›
          Gene Symbol:
          HFE
          Gene Name:
          hemochromatosis
          Gene Aliases:
          HFE1, HH, HLA-H, MVCD7, TFQTL2
          Chromosome Location:
          Chr.6: 26087281 - 26096216 on Build GRCh38
          UniGene ID:
          Hs.233325
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536592

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HFE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000410.3 NP_000401.1 5, 6 1161
          NM_139003.2 NP_620572.1 4, 5 843
          NM_139004.2 NP_620573.1 4, 5 885
          NM_139006.2 NP_620575.1 5, 6 1119
          NM_139007.2 NP_620576.1 4, 5 897
          NM_139008.2 NP_620577.1 4, 5 855
          NM_139009.2 NP_620578.1 5, 6 1092
          NM_139010.2 NP_620579.1 3, 4 621
          NM_139011.2 NP_620580.1 2, 3 345
          XM_011514543.2 XP_011512845.1 5, 6 1123
          GenBank mRNA AF079407.1 - 5, 6 995
          AF079408.1 - 4, 5 773
          AF079409.1 - 4, 5 731
          AF115265.1 - 5, 6 1012
          AF144242.1 - 4, 5 848
          AF149804.1 - 4, 5 727
          AJ249335.1 - 5, 6 1092
          AJ249336.1 - 4, 5 897
          AJ249337.1 - 4, 5 885
          AJ249338.1 - 2, 3 345
          AJ250635.1 - 3, 4 621
          AY205604.1 - 4, 5 762
          BC074721.2 - 6, 7 1041 30915171
          BC117201.1 - 5, 6 1036 40125752
          BC117203.1 - 5, 6 1036 40125754
          JN375755.1 - 2, 3 219
          JN375756.1 - 2, 3 218
          JN375757.1 - 2, 3 209
          JN375758.1 - 2, 3 218
          JN375759.1 - 2, 3 224
          U60319.1 - 5, 6 1222

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          negative regulation of T cell antigen processing and presentation
          negative regulation of T cell cytokine production
          protein complex assembly
          cellular iron ion homeostasis
          acute-phase response
          immune response
          female pregnancy
          response to iron ion
          cellular response to iron ion starvation
          positive regulation of gene expression
          positive regulation of pathway-restricted SMAD protein phosphorylation
          antigen processing and presentation
          BMP signaling pathway
          positive regulation of protein binding
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          hormone biosynthetic process
          positive regulation of receptor-mediated endocytosis
          iron ion homeostasis
          multicellular organismal iron ion homeostasis
          cellular response to iron ion
          positive regulation of peptide hormone secretion
          liver regeneration
          iron ion import into cell
          negative regulation of receptor binding
          positive regulation of receptor binding
          positive regulation of ferrous iron import into cell
          negative regulation of antigen processing and presentation of endogenous peptide antigen via MHC class I
          positive regulation of ferrous iron binding
          positive regulation of transferrin receptor binding
          response to iron ion starvation
          regulation of protein localization to cell surface
          negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of receptor activity
          positive regulation of receptor activity
          negative regulation of CD8-positive, alpha-beta T cell activation
          antigen binding
          receptor binding
          protein binding
          beta-2-microglobulin binding
          co-receptor binding
          peptide antigen binding
          transferrin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0