Search
Search

GALNTL6| Gene Symbol: GALNTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034845.2 | NP_001030017.2 | 7 | 1520 | ||
| XM_011531993.2 | XP_011530295.1 | 6 | 858 | |||
| XM_011531994.1 | XP_011530296.1 | 6 | 692 | |||
| XM_011531995.1 | XP_011530297.1 | 5 | 603 | |||
| XM_011531996.1 | XP_011530298.1 | 4 | 503 | |||
| XM_011531997.1 | XP_011530299.1 | 4 | 425 | |||
| XM_017008243.1 | XP_016863732.1 | 7 | 2254 | |||
| XM_017008244.1 | XP_016863733.1 | 6 | 1626 | |||
| GenBank mRNA | AJ626725.1 | - | 6 | 733 | ||
| AK295464.1 | - | 6 | 936 | |||
| BC047551.1 | - | 7 | 1441 | 5301536 | ||
| GU220060.1 | - | 6 | 784 | |||