Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD36B SIRNA Assays ›
          Gene Symbol:
          ANKRD36B ANKRD36 ANKRD36C
          Gene Name:
          ankyrin repeat domain 36B
          ankyrin repeat domain 36
          ankyrin repeat domain 36C
          Gene Aliases:
          KIAA1641
          UNQ2430
          -
          Chromosome Location:
          Chr.2: 97492663 - 97590245 on Build GRCh38
          Chromosome Location:
          Chr.2: 97113186 - 97264521 on Build GRCh38
          Chromosome Location:
          Chr.2: 95849548 - 95992320 on Build GRCh38
          UniGene ID:
          Hs.532921
          Hs.541894
          Hs.646318
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s60962

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD36B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_025190.3 NP_079466.3 38 3050
          XM_005263995.4 XP_005264052.1 38 3330
          XM_006712661.1 XP_006712724.1 38 3330
          XM_017004578.1 XP_016860067.1 38 3327
          XM_017004579.1 XP_016860068.1 36 3228
          XM_017004580.1 XP_016860069.1 36 3228
          XM_017004581.1 XP_016860070.1 36 3228
          XM_017004582.1 XP_016860071.1 36 3228
          XM_017004583.1 XP_016860072.1 37 3192
          XM_017004584.1 XP_016860073.1 32 3024
          XM_017004585.1 XP_016860074.1 36 3085
          XM_017004586.1 XP_016860075.1 33 2071
          XM_017004587.1 XP_016860076.1 37 3259
          XM_017004588.1 XP_016860077.1 36 3124
          XM_017004589.1 XP_016860078.1 35 3017
          XM_017004590.1 XP_016860079.1 34 2944
          XM_017004591.1 XP_016860080.1 20 3048
          XM_017004592.1 XP_016860081.1 39 3374
          XM_017004593.1 XP_016860082.1 37 3180
          XM_017004594.1 XP_016860083.1 36 3073
          XM_017004595.1 XP_016860084.1 37 3129
          GenBank mRNA CA394381.1 - 3 426
          Gene Symbol: ANKRD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164315.1 NP_001157787.1 70 4778
          XM_006712514.3 XP_006712577.1 70 5010
          XM_011511130.1 XP_011509432.1 70 5010
          XM_017004009.1 XP_016859498.1 70 5010
          XM_017004010.1 XP_016859499.1 70 5007
          XM_017004011.1 XP_016859500.1 68 4908
          XM_017004012.1 XP_016859501.1 68 4908
          XM_017004013.1 XP_016859502.1 68 4908
          XM_017004014.1 XP_016859503.1 68 4908
          XM_017004015.1 XP_016859504.1 68 4908
          XM_017004016.1 XP_016859505.1 68 4908
          XM_017004017.1 XP_016859506.1 68 4908
          XM_017004018.1 XP_016859507.1 68 4908
          XM_017004019.1 XP_016859508.1 68 4908
          XM_017004020.1 XP_016859509.1 68 4908
          XM_017004021.1 XP_016859510.1 68 4908
          XM_017004022.1 XP_016859511.1 68 4908
          XM_017004023.1 XP_016859512.1 68 4908
          XM_017004024.1 XP_016859513.1 69 4872
          XM_017004025.1 XP_016859514.1 66 4806
          XM_017004026.1 XP_016859515.1 66 4806
          XM_017004027.1 XP_016859516.1 66 4806
          XM_017004028.1 XP_016859517.1 66 4806
          XM_017004029.1 XP_016859518.1 66 4806
          XM_017004030.1 XP_016859519.1 66 4806
          XM_017004031.1 XP_016859520.1 66 4806
          XM_017004032.1 XP_016859521.1 66 4806
          XM_017004033.1 XP_016859522.1 66 4806
          XM_017004034.1 XP_016859523.1 66 4806
          XM_017004035.1 XP_016859524.1 66 4806
          XM_017004036.1 XP_016859525.1 66 4806
          XM_017004037.1 XP_016859526.1 66 4806
          XM_017004038.1 XP_016859527.1 66 4806
          XM_017004039.1 XP_016859528.1 64 4704
          XM_017004040.1 XP_016859529.1 64 4704
          XM_017004041.1 XP_016859530.1 64 4704
          XM_017004042.1 XP_016859531.1 64 4704
          XM_017004043.1 XP_016859532.1 64 4704
          XM_017004044.1 XP_016859533.1 62 4602
          XM_017004045.1 XP_016859534.1 62 4602
          XM_017004046.1 XP_016859535.1 68 4765
          XM_017004047.1 XP_016859536.1 67 4697
          XM_017004048.1 XP_016859537.1 66 4624
          XM_017004049.1 XP_016859538.1 68 4799
          XM_017004050.1 XP_016859539.1 53 3203
          XM_017004051.1 XP_016859540.1 51 3417
          GenBank mRNA AB046861.1 - 12 1676
          AF543495.1 - 2 219
          BC146835.1 - 12 1687 8860491
          Gene Symbol: ANKRD36C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310154.1 NP_001297083.1 83 5755

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding
          ion channel inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0