Search
Search

Hpse| Gene Symbol: Hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152803.4 | NP_690016.1 | 11 | 1488 | ||
| GenBank mRNA | AF359507.1 | - | 11 | 1477 | ||
| AK040471.1 | - | 11 | 1489 | |||
| AK087283.1 | - | 11 | 1515 | |||
| AK154628.1 | - | 11 | 1489 | |||
| AY077467.1 | - | 11 | 1480 | |||
| AY151051.1 | - | 11 | 1399 | |||
| BC138782.1 | - | 11 | 1482 | 8861804 | ||
| BC138784.1 | - | 11 | 1482 | 8861806 | ||