Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LAPTM5 SIRNA Assays ›
          Gene Symbol:
          Laptm5
          Gene Name:
          lysosomal-associated protein transmembrane 5
          Gene Aliases:
          E3
          Chromosome Location:
          Chr.4: 130913334 - 130936148 on Build GRCm38
          UniGene ID:
          Mm.271868
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s69086

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Laptm5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010686.3 NP_034816.1 5, 6 527
          GenBank mRNA AF364050.1 - 5, 6 545
          AK031802.1 - 5, 6 541
          AK050633.1 - 5, 6 542
          AK089432.1 - 5, 6 577
          AK089508.1 - 5, 6 550
          AK089535.1 - 5, 6 542
          AK150299.1 - 5, 6 538
          AK152918.1 - 5, 6 538
          AK153877.1 - 5, 6 541
          AK155744.1 - 5, 6 535
          AK163146.1 - 5, 6 542
          AK170438.1 - 5, 6 529
          AK170690.1 - 5, 6 561
          AK170867.1 - 5, 6 545
          AK171053.1 - 5, 6 552
          AK171057.1 - 5, 6 577
          AK171105.1 - 5, 6 542
          AK171116.1 - 5, 6 542
          AK171149.1 - 5, 6 550
          AK171172.1 - 542
          AK171193.1 - 5, 6 542
          AK171213.1 - 5, 6 542
          AK171236.1 - 5, 6 542
          AK171244.1 - 5, 6 542
          AK171250.1 - 5, 6 563
          AK171269.1 - 5, 6 579
          AK171270.1 - 5, 6 545
          AK171274.1 - 5, 6 535
          AK171280.1 - 5, 6 575
          AK171285.1 - 5, 6 579
          AK171481.1 - 5, 6 578
          AK171562.1 - 5, 6 533
          AK171571.1 - 5, 6 559
          AK171575.1 - 5, 6 577
          AK172306.1 - 5, 6 542
          AK172486.1 - 5, 6 577
          AK172707.1 - 5, 6 542
          AK172717.1 - 2, 3 245
          AK172885.1 - 5, 6 517
          BC020993.1 - 5, 6 527 4037686
          BC055057.1 - 5, 6 514 6529760
          BC114403.1 - 5, 6 507
          BC158032.1 - 5, 6 723 9007335
          U29539.1 - 5, 6 545
          U51239.1 - 5, 6 566

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s)

          transport

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0