Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARHGEF2 SIRNA Assays ›
          Gene Symbol:
          Arhgef2
          Gene Name:
          rho/rac guanine nucleotide exchange factor (GEF) 2
          Gene Aliases:
          AA408978, GEF, GEF-H1, GEFH1, LFP40, Lbcl1, Lfc, P40, mKIAA0651
          Chromosome Location:
          Chr.3: 88593114 - 88649532 on Build GRCm38
          UniGene ID:
          Mm.239329
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s69103

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Arhgef2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198911.1 NP_001185840.1 15, 16 2061
          NM_001198912.1 NP_001185841.1 14, 15 1929
          NM_001198913.1 NP_001185842.1 14, 15 1923
          NM_008487.3 NP_032513.3 14, 15 1939
          GenBank mRNA AB093254.1 - 14, 15 2012
          AF177032.1 - 14, 15 1906
          AK144972.1 - 15, 16 2041
          AK156379.1 - 14, 15 1921
          AK158232.1 - 15, 16 2132
          AK167628.1 - 14, 15 1893
          AK171223.1 - 14, 15 1882
          BC006589.1 - 14, 15 1860 3587800

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001012079, XM_008761151, XM_006232654, XM_006232643, XM_006232648, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          cell morphogenesis
          immune system process
          actin filament organization
          negative regulation of microtubule depolymerization
          cell cycle
          mitotic nuclear division
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          regulation of Rho protein signal transduction
          intracellular signal transduction
          positive regulation of GTPase activity
          innate immune response
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of neurogenesis
          positive regulation of NF-kappaB transcription factor activity
          cell division
          cellular response to muramyl dipeptide
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          microtubule binding
          Rho GTPase binding
          Rac guanyl-nucleotide exchange factor activity
          metal ion binding
          Rac GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0