Search
Search

Tnfsf10| Gene Symbol: Tnfsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009425.2 | NP_033451.1 | 4 | 491 | ||
| GenBank mRNA | AK142960.1 | - | 4 | 493 | ||
| AK144931.1 | - | 508 | ||||
| AK153581.1 | - | 4 | 541 | |||
| AK157633.1 | - | 492 | ||||
| BC011469.1 | - | 4 | 457 | |||
| BC138807.1 | - | 4 | 465 | 8861829 | ||
| BC138808.1 | - | 4 | 465 | 8861830 | ||
| U37522.1 | - | 4 | 467 | |||