Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGAV SIRNA Assays ›
          Gene Symbol:
          ITGAV
          Gene Name:
          integrin subunit alpha V
          Gene Aliases:
          CD51, MSK8, VNRA, VTNR
          Chromosome Location:
          Chr.2: 186590063 - 186680902 on Build GRCh38
          UniGene ID:
          Hs.436873
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7568
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ITGAV
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144999.2 NP_001138471.1 18 1768
          NM_001145000.2 NP_001138472.1 16 1944
          NM_002210.4 NP_002201.1 18 2052
          GenBank mRNA AB209894.1 - 16 1927
          AK302990.1 - 18 1768
          BC126231.1 - 18 1905 8991947
          BC136442.1 - 18 1905 9020431
          BC144100.1 - 16 1797 9052620
          EF560727.1 - 18 1820
          M14648.1 - 18 1817

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00233808_m1

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          positive regulation of cell proliferation
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of lipid storage
          cell growth
          cell migration
          extracellular matrix organization
          positive regulation of cell migration
          cell-substrate adhesion
          negative regulation of lipid transport
          positive regulation of osteoblast proliferation
          heterotypic cell-cell adhesion
          substrate adhesion-dependent cell spreading
          endodermal cell differentiation
          apolipoprotein A-I-mediated signaling pathway
          apoptotic cell clearance
          negative regulation of low-density lipoprotein particle receptor biosynthetic process
          positive regulation of cell adhesion
          viral entry into host cell
          vascular endothelial growth factor receptor signaling pathway
          negative regulation of lipoprotein metabolic process
          regulation of phagocytosis
          leukocyte migration
          negative chemotaxis
          entry of symbiont into host cell by promotion of host phagocytosis
          ERK1 and ERK2 cascade
          calcium ion transmembrane transport
          extrinsic apoptotic signaling pathway in absence of ligand
          regulation of apoptotic cell clearance
          negative regulation of entry of bacterium into host cell
          negative regulation of extrinsic apoptotic signaling pathway
          virus receptor activity
          opsonin binding
          fibronectin binding
          protease binding
          protein kinase C binding
          voltage-gated calcium channel activity
          protein binding
          coreceptor activity
          fibroblast growth factor binding
          C-X3-C chemokine binding
          insulin-like growth factor I binding
          neuregulin binding
          metal ion binding
          transforming growth factor beta binding
          extracellular matrix binding
          extracellular matrix protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0