Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RHOA SIRNA Assays ›
          Gene Symbol:
          RHOA
          Gene Name:
          ras homolog family member A
          Gene Aliases:
          ARH12, ARHA, RHO12, RHOH12
          Chromosome Location:
          Chr.3: 49359136 - 49412097 on Build GRCh38
          UniGene ID:
          Hs.247077
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s760
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RHOA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001313941.1 NP_001300870.1 6 1364
          NM_001313943.1 NP_001300872.1 6 1408
          NM_001313944.1 NP_001300873.1 5 1209
          NM_001313945.1 NP_001300874.1 5 1146
          NM_001313946.1 NP_001300875.1 3 1017
          NM_001313947.1 NP_001300876.1 4 1148
          NM_001664.3 NP_001655.1 5 1269
          GenBank mRNA AK130066.1 - 5 1265
          AK130808.1 - 5 1145
          AK222556.1 - 5 1161
          BC000946.3 - 2 713
          BC001360.2 - 5 1101 3051460
          BC005976.1 - 5 1106 4102976
          BX647063.1 - 5 1165
          EL952052.1 - 1 184
          L09159.1 - 4 978
          L25080.1 - 5 1141

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00357608_m1

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          transforming growth factor beta receptor signaling pathway
          Rho protein signal transduction
          viral process
          cell migration
          substantia nigra development
          actin cytoskeleton organization
          platelet activation
          regulation of cell migration
          actin cytoskeleton reorganization
          positive regulation of cytokinesis
          regulation of actin cytoskeleton organization
          regulation of osteoblast proliferation
          Roundabout signaling pathway
          cleavage furrow formation
          apolipoprotein A-I-mediated signaling pathway
          positive regulation of NF-kappaB import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          stress fiber assembly
          apical junction assembly
          endothelial cell migration
          ossification involved in bone maturation
          wound healing, spreading of cells
          positive regulation of neuron differentiation
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          phosphatidylinositol-mediated signaling
          negative regulation of axonogenesis
          positive regulation of axonogenesis
          negative chemotaxis
          regulation of small GTPase mediated signal transduction
          positive regulation of stress fiber assembly
          Wnt signaling pathway, planar cell polarity pathway
          positive regulation of lipase activity
          trabecula morphogenesis
          positive regulation of protein serine/threonine kinase activity
          negative regulation of cell migration involved in sprouting angiogenesis
          mitotic spindle assembly
          endothelial tube lumen extension
          skeletal muscle satellite cell migration
          mitotic cleavage furrow formation
          regulation of cell motility
          GTPase activity
          protein binding
          GTP binding
          myosin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nbq2g:80/100.66.72.197:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0