Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253783.1 | NP_001240712.1 | 6 | 489 | ||
NM_001253784.1 | NP_001240713.1 | 5 | 467 | |||
NM_001281803.1 | NP_001268732.1 | 4 | 471 | |||
NM_054046.5 | NP_473387.1 | 4 | 380 | |||
GenBank mRNA | AK028726.1 | - | 5 | 439 | ||
AK034028.1 | - | 346 | ||||
AK046271.1 | - | 4 | 341 | |||
AK050405.1 | - | 5 | 438 | |||
AK086174.1 | - | 4 | 341 | |||
AK087778.1 | - | 4 | 351 | |||
AK129019.1 | - | 4 | 422 | |||
AK133979.1 | - | 4 | 354 | |||
AK150334.1 | - | 4 | 316 | |||
AK150994.1 | - | 4 | 316 | |||
AK164147.1 | - | 4 | 352 | |||
BC003306.1 | - | 6 | 440 | 3154712 | ||
BY713285.1 | - | 4 | 381 | |||
CX562217.1 | - | 4 | 433 |