Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012037.2 | NP_036167.1 | 6 | 1215 | ||
GenBank mRNA | AK034795.1 | - | 6 | 1217 | ||
AK145193.1 | - | 6 | 1249 | |||
AK154178.1 | - | 1300 | ||||
AK154965.1 | - | 6 | 1198 | |||
AK155342.1 | - | 6 | 1217 | |||
AK159310.1 | - | 6 | 1158 | |||
AK159315.1 | - | 6 | 1200 | |||
AK160891.1 | - | 6 | 1207 | |||
AK170371.1 | - | 6 | 1196 | |||
AK216339.1 | - | 2 | 204 | |||
BC024586.1 | - | 4 | 444 | |||
BC025496.1 | - | 6 | 1171 | 5320239 | ||
BC036169.1 | - | 5 | 657 | |||
BC085504.1 | - | 6 | 1134 | |||
BC099678.1 | - | 6 | 1197 | 6311681 |