Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012037.2 | NP_036167.1 | 4 | 949 | ||
GenBank mRNA | AK034795.1 | - | 4 | 951 | ||
AK145193.1 | - | 4 | 983 | |||
AK154178.1 | - | 934 | ||||
AK154965.1 | - | 4 | 932 | |||
AK155342.1 | - | 4 | 951 | |||
AK159310.1 | - | 4 | 892 | |||
AK159315.1 | - | 4 | 934 | |||
AK160891.1 | - | 4 | 941 | |||
AK170371.1 | - | 4 | 930 | |||
AK178348.1 | - | 2 | 203 | |||
AK181513.1 | - | 2 | 203 | |||
AK183725.1 | - | 2 | 203 | |||
AK185056.1 | - | 3 | 283 | |||
BC024586.1 | - | 2 | 178 | |||
BC025496.1 | - | 4 | 905 | 5320239 | ||
BC036169.1 | - | 3 | 391 | |||
BC085504.1 | - | 4 | 868 | |||
BC099678.1 | - | 4 | 931 | 6311681 |