Search
Search

KPNA5| Gene Symbol: KPNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002269.2 | NP_002260.2 | 12 | 1316 | ||
| XM_011535805.1 | XP_011534107.1 | 13 | 1494 | |||
| XM_017010838.1 | XP_016866327.1 | 13 | 1494 | |||
| XM_017010839.1 | XP_016866328.1 | 12 | 1297 | |||
| XM_017010840.1 | XP_016866329.1 | 13 | 1454 | |||
| XM_017010841.1 | XP_016866330.1 | 9 | 877 | |||
| XM_017010842.1 | XP_016866331.1 | 11 | 1208 | |||
| XM_017010843.1 | XP_016866332.1 | 12 | 1332 | |||
| GenBank mRNA | AF005361.1 | - | 11 | 1181 | ||
| AK314206.1 | - | 12 | 1309 | |||
| BC047409.1 | - | 12 | 1308 | 5297277 | ||