Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CADM1 SIRNA Assays ›
          Gene Symbol:
          Cadm1
          Gene Name:
          cell adhesion molecule 1
          Gene Aliases:
          2900073G06Rik, 3100001I08Rik, AI987920, Bl2, Igsf4, Igsf4a, Necl2, RA175, RA175A, RA175B, RA175C, RA175N, ST17, SgIGSF, SynCam, Tslc1
          Chromosome Location:
          Chr.9: 47530239 - 47853385 on Build GRCm38
          UniGene ID:
          Mm.234832
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s79874

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cadm1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025600.1 NP_001020771.1 2 169
          NM_018770.3 NP_061240.3 2 169
          NM_207675.2 NP_997558.2 2 169
          NM_207676.2 NP_997559.1 2 169
          GenBank mRNA AB052293.1 - 2 169
          AB064265.2 - 2 148
          AB092414.1 - 2 148
          AB183399.1 - 2 148
          AB183400.1 - 2 148
          AB183401.1 - 2 148
          AB183402.1 - 2 148
          AB377192.1 - 2 169
          AF434663.1 - 2 264
          AF539424.1 - 2 161
          AK013775.1 - 2 170
          AK013911.1 - 2 263
          AK053839.1 - 2 170
          AK172628.1 - 2 172
          AY351388.1 - 2 148
          BC057125.1 - 2 157 6414884
          BC090970.1 - 2 348 6703838
          BC095986.1 - 2 161 5372277
          DQ279856.1 - 2 148
          JQ782219.1 - 2 148

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          immune system process
          apoptotic process
          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          multicellular organism development
          spermatogenesis
          synapse assembly
          cell recognition
          unidimensional cell growth
          calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
          cell differentiation
          susceptibility to natural killer cell mediated cytotoxicity
          positive regulation of natural killer cell mediated cytotoxicity
          positive regulation of cytokine secretion
          detection of stimulus
          bone development
          receptor binding
          protein binding
          PDZ domain binding
          protein homodimerization activity
          cell adhesion molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0