Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAGEA3 SIRNA Assays ›
          Gene Symbol:
          MAGEA3 MAGEA2 MAGEA2B MAGEA6
          Gene Name:
          MAGE family member A3
          MAGE family member A2
          MAGE family member A2B
          MAGE family member A6
          Gene Aliases:
          CT1.3, HIP8, HYPD, MAGE3, MAGEA6
          CT1.2, MAGE2, MAGEA2A
          CT1.2, MAGE2
          CT1.6, MAGE-3b, MAGE3B, MAGE6
          Chromosome Location:
          Chr.X: 152698742 - 152702347 on Build GRCh38
          Chromosome Location:
          Chr.X: 152749863 - 152753942 on Build GRCh38
          Chromosome Location:
          Chr.X: 152714529 - 152718607 on Build GRCh38
          Chromosome Location:
          Chr.X: 152766136 - 152769729 on Build GRCh38
          UniGene ID:
          Hs.417816
          Hs.534597
          Hs.534597
          Hs.441113
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s8437

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAGEA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005362.3 NP_005353.1 3 1067
          XM_005274676.3 XP_005274733.1 3 1141
          XM_006724818.2 XP_006724881.1 3 1223
          XM_011531160.2 XP_011529462.1 3 1047
          XM_011531161.2 XP_011529463.1 3 1472
          GenBank mRNA AK223519.1 - 1 894
          AK292384.1 - 3 1067
          BC000340.2 - 3 1032 2822978
          BC005963.1 - 3 1067 4064879
          BC011744.2 - 2 993 3345801
          BC016803.1 - 3 1029 4073332
          BC017389.1 - 3 1024 4047542
          CR541767.1 - 1 858
          CR541768.1 - 1 858
          CR541774.1 - 1 858
          CR542020.1 - 1 858
          Gene Symbol: MAGEA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282501.1 NP_001269430.1 5 1304
          NM_001282502.1 NP_001269431.1 4 1206
          NM_001282504.1 NP_001269433.1 3 1086
          NM_001282505.1 NP_001269434.1 2 1020
          NM_005361.3 NP_005352.1 4 1381
          NM_175742.2 NP_786884.1 6 1367
          NM_175743.2 NP_786885.1 6 1383
          XM_017029519.1 XP_016885008.1 5 1343
          GenBank mRNA AK290443.1 - 5 1304
          BC013098.2 - 2 990 4343986
          BC063681.1 - 4 1170 4403536
          BC108720.1 - 4 1200 6065905
          BC112158.1 - 1 888 8327626
          BC112160.1 - 1 888 8327628
          Gene Symbol: MAGEA2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321400.1 NP_001308329.1 6 1383
          NM_001321401.1 NP_001308330.1 6 1367
          NM_001321402.1 NP_001308331.1 5 1304
          NM_001321403.1 NP_001308332.1 4 1206
          NM_001321404.1 NP_001308333.1 2 1020
          NM_153488.4 NP_705692.1 3 1086
          XM_017029406.1 XP_016884895.1 5 1342
          Gene Symbol: MAGEA6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005363.3 NP_005354.1 3 1065
          NM_175868.2 NP_787064.1 3 1129
          GenBank mRNA AK289437.1 - 3 1049
          BC041599.1 - 3 1038 5458512
          BC067731.1 - 3 1046 4828429
          BX640600.1 - 3 1426
          D32076.1 - 1 923
          EU161101.1 - 1 858
          U10339.1 - 1 858

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein processing
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          negative regulation of protein sumoylation
          cellular protein catabolic process
          positive regulation of ubiquitin-protein transferase activity
          signal transduction by p53 class mediator
          cellular senescence
          negative regulation of protein acetylation
          biological_process
          protein binding
          caspase binding
          ubiquitin protein ligase binding
          histone deacetylase binding
          repressing transcription factor binding
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nglt7:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0