Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027999.1 | NP_082275.1 | 18 | 1783 | ||
GenBank mRNA | AK009476.1 | - | 18 | 1763 | ||
AK129228.1 | - | 14 | 3051 | |||
AK153920.1 | - | 18 | 1766 | |||
BC023723.1 | - | 18 | 1672 | |||
BC048724.1 | - | 4 | 408 | |||
BC089002.1 | - | 18 | 1752 | |||
BC118934.1 | - | 18 | 1758 | 40090953 | ||
BC118935.1 | - | 18 | 1758 | 40090954 |