Search
Search

Glt8d1| Gene Symbol: Glt8d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165930.1 | NP_001159402.1 | 9 | 938 | ||
| NM_029626.2 | NP_083902.2 | 9 | 1467 | |||
| GenBank mRNA | AK010388.1 | - | 9 | 1196 | ||
| AK077388.1 | - | 9 | 931 | |||
| AK217645.1 | - | 3 | 237 | |||
| BC029215.1 | - | 7 | 1139 | |||
| BC055709.1 | - | 8 | 1354 | |||
| BC058731.1 | - | 8 | 1354 | |||
| BC069873.1 | - | 9 | 1017 | 6314675 | ||