Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB089793.1 | - | 9 | 746 | ||
AF482993.1 | - | 8 | 710 | |||
AK049410.1 | - | 9 | 742 | |||
AK051791.1 | - | 8 | 661 | |||
AK143281.1 | - | 9 | 715 | |||
AK160826.1 | - | 7 | 584 | |||
AK173069.1 | - | 9 | 1082 | |||
AK216559.1 | - | 2 | 114 | |||
BC030879.1 | - | 9 | 745 | 4241988 | ||
BC034508.1 | - | 2 | 146 | |||
BC058947.1 | - | 9 | 716 | 5702781 |