Search
Search

S1pr5| Gene Symbol: S1pr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053190.2 | NP_444420.1 | 3 | 920 | ||
| GenBank mRNA | AF327535.1 | - | 1 | 599 | ||
| AK081126.1 | - | 1 | 599 | |||
| BC012232.1 | - | 2 | 845 | 4194177 | ||
Set Membership: |
|