Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159908.1 | NP_001153380.1 | 4 | 464 | ||
NM_133349.3 | NP_579927.1 | 4 | 475 | |||
GenBank mRNA | AF224494.1 | - | 4 | 361 | ||
AK146082.1 | - | 4 | 413 | |||
AK171245.1 | - | 4 | 415 | |||
AK198290.1 | - | 2 | 92 | |||
BC058780.1 | - | 4 | 401 | 6312202 |