Search
Search

Egln3| Gene Symbol: Egln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028133.2 | NP_082409.2 | 3 | 891 | ||
| GenBank mRNA | AJ310548.1 | - | 3 | 872 | ||
| AK011502.1 | - | 2 | 195 | |||
| AK044787.1 | - | 3 | 893 | |||
| AK165972.1 | - | 3 | 822 | |||
| AK170732.1 | - | 3 | 822 | |||
| BC044926.1 | - | 3 | 801 | 5373240 | ||
| BC058278.1 | - | 3 | 811 | 6528802 | ||
| BC069893.1 | - | 3 | 819 | 4978765 | ||